Skip to Content
Merck
All Photos(1)

Key Documents

76245

Sigma-Aldrich

Atto 655 NHS ester

BioReagent, suitable for fluorescence

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

Assay

≥90.0% (HPLC)

form

powder

manufacturer/tradename

ATTO-TEC GmbH

extent of labeling

≥90.0%

λ

in ethanol (with 0.1% trifluoroacetic acid)

UV absorption

λ: 650.0-656.0 nm Amax

suitability

suitable for coupling to amines
suitable for fluorescence

storage temp.

−20°C

SMILES string

[S](=O)(=O)([O-])CC1CC([N+](=C3C1=CC4=Nc5c(cc6c(c5)CCCN6CC)OC4=C3)CCCC(=O)ON2C(=O)CCC2=O)(C)C

InChI

1S/C31H36N4O8S/c1-4-33-11-5-7-19-13-22-26(15-24(19)33)42-27-16-25-21(14-23(27)32-22)20(18-44(39,40)41)17-31(2,3)34(25)12-6-8-30(38)43-35-28(36)9-10-29(35)37/h13-16,20H,4-12,17-18H2,1-3H3

InChI key

SPDKWNQUMMQOKX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Atto fluorescent labels are designed for high sensitivity applications, including single molecule detection. Atto labels have rigid structures that do not show any cis-trans-isomerization. Thus these labels display exceptional intensity with minimal spectral shift on conjugation.

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gerald Giller et al.
Nucleic acids research, 31(10), 2630-2635 (2003-05-09)
Fluorescent-labeled DNA is generated through enzymatic incorporation of fluorophore-linked 2'-deoxyribonucleoside-5'-triphosphates (dNTPs) by DNA polymerases. We describe the synthesis of a variety of dye-labeled dNTPs. Amino-linker-modified 5'-triphosphates of all four naturally occurring nucleobases were used as precursors. Commercially available dyes were
Highly sensitive protease assay using fluorescence quenching of peptide probes based on photoinduced electron transfer.
Nicole Marmé et al.
Angewandte Chemie (International ed. in English), 43(29), 3798-3801 (2004-07-20)
Ye Chen et al.
Biomedical optics express, 10(3), 1257-1272 (2019-03-21)
Open-top light-sheet microscopy is a technique that can potentially enable rapid ex vivo inspection of large tissue surfaces and volumes. Here, we have optimized an open-top light-sheet (OTLS) microscope and image-processing workflow for the comprehensive examination of surgical margin surfaces
Taurai Tasara et al.
Nucleic acids research, 31(10), 2636-2646 (2003-05-09)
The modification of nucleic acids using nucleotides linked to detectable reporter or functional groups is an important experimental tool in modern molecular biology. This enhances DNA or RNA detection as well as expanding the catalytic repertoire of nucleic acids. Here
Nicole Marmé et al.
Bioconjugate chemistry, 14(6), 1133-1139 (2003-11-20)
Steady-state and time-resolved fluorescence measurements were performed to elucidate the fluorescence quenching of oxazine, rhodamine, carbocyanine, and bora-diaza-indacene dyes by amino acids. Among the natural amino acids, tryptophan exhibits the most pronounced quenching efficiency. Especially, the red-absorbing dyes ATTO 655

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service