Skip to Content
Merck
All Photos(4)

Key Documents

F2456

Sigma-Aldrich

Fluorescein (free acid)

Fluorescein (free acid)
1 of 1 reviewers received a sample product or took part in a promotion

Dye content 95%, Powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H12O5
CAS Number:
Molecular Weight:
332.31
Colour Index Number:
45350:1
Beilstein:
94324
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
Pricing and availability is not currently available.

Product Name

Fluorescein (free acid), Dye content 95 %

form

powder

composition

Dye content, 95%

technique(s)

titration: suitable

color

orange to red-brown

mp

320 °C (lit.)

λmax

496 nm

ε (extinction coefficient)

≥12000 at 282-286 nm
≥39000 at 236-240 nm
≥70000 at 488-492 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Oc1ccc2c(Oc3cc(O)ccc3C24OC(=O)c5ccccc45)c1

InChI

1S/C20H12O5/c21-11-5-7-15-17(9-11)24-18-10-12(22)6-8-16(18)20(15)14-4-2-1-3-13(14)19(23)25-20/h1-10,21-22H

InChI key

GNBHRKFJIUUOQI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fluorescein (free acid, CI 45350) is a xanthene dye.[1] Fluorescein (free acid) molecule can be divided into two parts. One part contains xanthene moiety which acts as fluorophore and the other part of the molecule bears benzene moiety which is the fluorescence-controlling moiety.

Application

Fluorescein (free acid) has been used as a staining agent in retinal imaging.[2]
Fluorescein (free acid) is widely used as a fluorescent stain and trace for many living cells and tissue applications. It is also suitable as a marker for the study of motility of bile canaliculi. Some of the clinical applications are evaluating the ocular surface and delineation of brain regions lacking a blood-brain barrier to permit accurate surgical resection of malignant glioma. Fluorescein angiography helps investigate changes in retinal blood supply. Fluorescein also plays a role as a physiological tracer to characterize the salmon′s intestinal permeability (Schep et al., 1998), and for the fluorophotometric measurement of aqueous humor flow. It is widely employed as a quantum yield standard.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karen A Fisher et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 9(23), 5873-5878 (2003-12-16)
The uptake of the three species of the drug model fluorescein (fluorescein sodium salt (FNa), fluorescein free acid (F), and fluorescein diacetate (FDA)) by zeolite NaX and the mesoporous zeotype MCM-41 was investigated as well as their release rates into
Cunyang Wang et al.
Materials science & engineering. C, Materials for biological applications, 103, 109823-109823 (2019-07-28)
Acrylic bone cement (ABC) has been used as a grouting agent in joint replacement surgery for over 50 years. In particular, ABC is irreplaceable for high-load joint replacement such as total hip joint replacements (THJRs) and total knee joint replacements because
Han Liu et al.
Journal of ethnopharmacology, 241, 111975-111975 (2019-05-30)
Danhong Huayu Koufuye (DHK), a compound traditional Chinese medicine, is composed of Salvia miltiorrhiza radix (Salvia miltiorrhiza Bge.), Angelicae Sinensis radix (Angelicae Sinensis (Oliv.) Diels.), Chuanxiong rhizoma (Ligusticum chuanxiong Hort.), Persicae semen (Prunus persica (L.) Batsch), Carthami flos (Carthamus tinctorius
Tasuku Ueno et al.
Journal of the American Chemical Society, 126(43), 14079-14085 (2004-10-28)
Rational design strategies based on practical fluorescence modulation mechanisms would enable us to rapidly develop novel fluorescence probes for target molecules. Here, we present a practical and general principle for modulating the fluorescence properties of fluorescein. We hypothesized that (a)
Mi Ju Son et al.
Medicine, 98(29), e16527-e16527 (2019-07-25)
Several studies have found that obesity is associated with atopic dermatitis (AD); however, the mechanisms underlying the association are largely unknown. This study aims to assess the association of AD with obesity in the Korean population and verify its mechanism

Questions

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. Brown University Providence RI
    • Review 1
    • Votes 4
    1 out of 5 stars.

    new product not working when the old one did

    This product didn't work. i was wondering if we could be a credit or reimbursed?
    We have used this in the last (lot# BCCD8787) and it worked but the new one we got (BCCH4248) didn't.

    Helpful?

    1. Response from MilliporeSigma:

      Thank you for taking the time to leave a review. We are sorry to hear that your experience did not meet expectations. Our Customer Service team has reached out to you in effort to resolve this issue. We apologize for any inconvenience this may have caused.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service