Skip to Content
Merck
All Photos(1)

Key Documents

53796

Sigma-Aldrich

(Z-)-4-Bromo-5-(bromomethylene)-2(5H)-furanone

≥97.0% (GC)

Synonym(s):

Furanone C-30

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H2Br2O2
CAS Number:
Molecular Weight:
253.88
Beilstein:
120928
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥97.0% (GC)
97.0-103.0% (T)

form

solid

storage temp.

−20°C

SMILES string

Br\C=C1/OC(=O)C=C1Br

InChI

1S/C5H2Br2O2/c6-2-4-3(7)1-5(8)9-4/h1-2H/b4-2-

InChI key

DPGLBHQUHFJRJS-RQOWECAXSA-N

Biochem/physiol Actions

The synthetic furanones C30 interfered with the actions of acyl-HSL molecules and inhibited gene expression regulated by quorum-sensing systems in P. aeruginosa.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jingming Zhao et al.
The Indian journal of medical research, 147(4), 400-406 (2018-07-13)
Pseudomonas aeruginosa is an opportunistic pathogen that can cause nosocomial bloodstream infections in humans. This study was aimed to explore the association of furanone C-30 with biofilm formation, quorum sensing (QS) system and antibiotic resistance in P. aeruginosa. An in
Young Mok Heo et al.
Marine drugs, 16(12) (2018-12-19)
In our previous study, all Arthrinium isolates from Sargassum sp. showed high bioactivities, but studies on marine Arthrinium spp. are insufficient. In this study, a phylogenetic analysis of 28 Arthrinium isolates from seaweeds and egg masses of Arctoscopus japonicus was
J Fernando Montes García et al.
Antonie van Leeuwenhoek, 113(9), 1371-1383 (2020-07-17)
Actinobacillus seminis is an autochthonous gram-negative bacterium that affects reproductive organs, causing epididymitis, low fertility, and occasional abortions in ovine and goats. The virulence factors and the pathogenicity mechanisms of A. seminis have not been clearly elucidated yet. In this
Kulwadee Karnjana et al.
Fish & shellfish immunology, 88, 91-101 (2019-03-01)
Bacteria respond to host immunity for their proliferation and survival by cell-cell communications such as biofilm formation, bioluminescence, and secreting virulence factors. In the biofilm form, bacteria are more resistant to various antimicrobial treatments and withstand the host's immune system.
B X V Quecan et al.
Frontiers in microbiology, 10, 867-867 (2019-05-21)
Quorum sensing (QS) regulates bacterial gene expression and studies suggest quercetin, a flavonol found in onion, as a QS inhibitor. There are no studies showing the anti-QS activity of plants containing quercetin in its native glycosylated forms. This study aimed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service