Skip to Content
Merck
All Photos(4)

Key Documents

O3379

Sigma-Aldrich

1-Octacosanol

≥99% (GC)

Synonym(s):

n-Octacosanol, Octacosanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H58O
CAS Number:
Molecular Weight:
410.76
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

Assay

≥99% (GC)

form

powder

lipid type

oils

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCCCCCO

InChI

1S/C28H58O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26-27-28-29/h29H,2-28H2,1H3

InChI key

CNNRPFQICPFDPO-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tao Wang et al.
Acta pharmacologica Sinica, 31(7), 765-774 (2010-06-29)
To investigate the protective effects of octacosanol in 6-hydroxydopamine-induced Parkinsonian rats and find whether octacosanol has effects on pro nerve growth factor (pro-NGF), NGF and the downstream effector proteins. Behavioral tests, enzymatic assay, tyrosine hydroxylase immunohistochemistry, TUNEL and Western blot
Mahesh K Kaushik et al.
Scientific reports, 7(1), 8892-8892 (2017-08-23)
Octacosanol, a component of various food materials, possesses prominent biological activities and functions. It fights against cellular stress by increasing glutathione level and thus scavenging oxygen reactive species. However, its anti-stress activity and role in sleep induction remained elusive. We
Christopher P F Marinangeli et al.
The British journal of nutrition, 97(2), 381-388 (2007-02-15)
Policosanols (PC) exist as very-long-chain alcohols derived from sugarcane currently used in many countries as a cholesterol-lowering therapy. PC purity and relative percentage composition have been suggested as primary reasons why the original Cuban PC (OPC) supplements possess lipid-lowering efficacy.
Amira N Kassis et al.
Atherosclerosis, 194(1), 153-158 (2006-11-25)
Policosanol is a mixture of higher aliphatic alcohols shown to have beneficial effects on plasma lipid levels in animals and humans. Over 50 studies have reported significant reductions in plasma cholesterol using policosanol obtained from Cuban sugar cane (Dalmer, La
Arrigo F G Cicero et al.
Complementary therapies in medicine, 13(4), 273-278 (2005-12-13)
We planned to carry out a pilot study to evaluate the efficacy and safety as an antihypercholesterolemic agent of a brand dietary supplement made of Monascus purpureus titrated extract, octacosanols and niacin on 111 Caucasian patients with low cardiovascular disease

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service