Skip to Content
Merck
All Photos(1)

Key Documents

42052

Sigma-Aldrich

cis-11-Methyl-2-dodecenoic acid

≥90.0% (HPLC)

Synonym(s):

DSF, Diffusible signal factor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H24O2
CAS Number:
Molecular Weight:
212.33
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥90.0% (HPLC)

storage temp.

−20°C

SMILES string

CC(C)CCCCCCC\C=C/C(O)=O

InChI

1S/C13H24O2/c1-12(2)10-8-6-4-3-5-7-9-11-13(14)15/h9,11-12H,3-8,10H2,1-2H3,(H,14,15)/b11-9-

InChI key

SNTXNGAQYNSTHI-LUAWRHEFSA-N

Application

Diffusible signal factor (cis-11-methyl-2-dodecenoic acid, DSF) is used to study cell-cell communication by pathogenic bacteria such as Xanthomonas campestris pv. Campestris (Xcc). DSF is involved in the synchronization of virulence gene expression and biofilm dispersal.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Ben E Clifton et al.
Nature chemical biology, 14(6), 542-547 (2018-04-25)
The emergence of enzymes through the neofunctionalization of noncatalytic proteins is ultimately responsible for the extraordinary range of biological catalysts observed in nature. Although the evolution of some enzymes from binding proteins can be inferred by homology, we have a
Yaping Yang et al.
Scanning, 38(6), 825-836 (2016-06-30)
Disulfiram (DSF) has been proved to have broad-spectrum anti-alcoholism effects, and it is also found to show stronger anti-tumor effects after chelating with Cu2+ to form DSF-Cu complex. In this work, we studied the anti-tumor activity of DSF-Cu in MCF-7
Abeer A Ageeli et al.
Nucleic acids research, 47(3), 1468-1481 (2018-11-22)
Nucleic acid triplexes may regulate many important biological processes. Persistent accumulation of the oncogenic 7-kb long noncoding RNA MALAT1 is dependent on an unusually long intramolecular triple helix. This triplex structure is positioned within a conserved ENE (element for nuclear
Anita Sarkar et al.
Nature communications, 9(1), 1956-1956 (2018-05-18)
Furin cleavage of the HIV envelope glycoprotein is an essential step for cell entry that enables formation of well-folded, native-like glycosylated trimers, releases constraints on the fusion peptide, and limits enzymatic processing of the N-glycan shield. Here, we show that
Kaku Goto et al.
Oncotarget, 7(46), 74987-74999 (2016-09-08)
We recently described that the anti-apoptotic AMPK-related kinase, SNARK, promotes transforming growth factor (TGF)-β signaling in hepatocellular carcinoma (HCC) cells, as a potentially new therapeutic target. Here we explored FDA-approved drugs inhibiting the enzymatic activity of SNARK, using an in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service