Skip to Content
Merck
All Photos(2)

Key Documents

1801

Sigma-Aldrich

α-Naphthyl Butyrate Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116124
NACRES:
NA.47

form

solution

IVD

for in vitro diagnostic use

concentration

2.4 g/L

application(s)

hematology
histology

shipped in

dry ice

storage temp.

−20°C

Application

Intended for use in the Sigma α-Butyrate Esterase kit, procedure 181B

Packaging

2.4g/L alpha-naphthyl butyrate in methanol solution with stabilizers

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3 - STOT SE 1

Target Organs

Eyes,Central nervous system

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel J Liebertz et al.
Head & neck oncology, 2, 5-5 (2010-02-24)
Head and neck squamous cell carcinoma (HNSCC) is an aggressive and lethal malignancy. Publically available cell lines are mostly of lingual origin, or have not been carefully characterized. Detailed characterization of novel HNSCC cell lines is needed in order to
Marta Sanchez-Carbayo et al.
The American journal of pathology, 163(2), 505-516 (2003-07-24)
To identify gene expression changes along progression of bladder cancer, we compared the expression profiles of early-stage and advanced bladder tumors using cDNA microarrays containing 17,842 known genes and expressed sequence tags. The application of bootstrapping techniques to hierarchical clustering
Axel Hoos et al.
The American journal of pathology, 160(1), 175-183 (2002-01-12)
Hürthle cell tumors are rare thyroid neoplasms for which disease biology is poorly understood and diagnosis of carcinoma can be challenging. The aim of the study was to characterize molecular expression profiles of Hürthle cell tumors and to determine the
Vijay G Ramakrishnan et al.
Journal of biomolecular techniques : JBT, 16(4), 297-305 (2006-03-09)
Ticks are obligate ectoparasites that feed on a variety of hosts including mammals, birds and reptiles. Prolonged attachment on the host and an ability to transmit a wide variety of pathogens are the special features of tick feeding. Salivary glands
Michael D'Angelica et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 24(9), 1221-1228 (2011-05-17)
Folate receptor alpha (FRα), encoded by folate receptor 1 (adult) gene, has emerged as a cancer biomarker and potential therapeutic target. In addition, its expression in tumors may offer prognostic information. The aim of this study was to assess the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service