N8505
1-Naphthyl acetate
≥98% (C)
Synonym(s):
α-Naphthyl acetate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3CO2C10H7
CAS Number:
Molecular Weight:
186.21
Beilstein:
2046403
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77
Recommended Products
Assay
≥98% (C)
form
crystals
mp
43-46 °C (lit.)
storage temp.
−20°C
SMILES string
CC(=O)Oc1cccc2ccccc12
InChI
1S/C12H10O2/c1-9(13)14-12-8-4-6-10-5-2-3-7-11(10)12/h2-8H,1H3
InChI key
VGKONPUVOVVNSU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
1-Naphthyl acetate is a 1-napthyl ester. Mechanism of photo-Fries rearrangement of 1-naphthyl acetate has been studied using steady state photolysis and laser flash photolysis. Kinetics of photo-Fries rearrangement of 1-napthyl acetate has been studied using steady-state, time-resolved chemically induced dynamic nuclear polarization (CIDNP) and flash photolysis methods.
Application
1-Naphthyl acetate may be used in a rapid staining method for identification of macrophages; counts by this method were confirmed by the more complex morphological criteria, by phagocytosis, and by the presence of Fc receptors. It may be employed as substrate to investigate the distribution of non-specific carboxylic esterases (EC 3.1.1) in the digestive tract of perch, Perm fliiviatilis L.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
235.4 °F
Flash Point(C)
113 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Magnetic isotope and external magnetic field effects upon the photo-Fries rearrangement of 1-naphthyl acetate.
Nakagaki R, et al.
The Journal of Physical Chemistry, 89(15), 3222-3226 (1985)
Non-specific carboxylic esterase activity in the digestive tract of the perch, Perca fluviatilis L.
Hirji KN and Courtney WAM.
Journal of Fish Biology, 22(1), 1-7 (1983)
Purification and kinetic characterisation of juvenile hormone esterase from Drosophila melanogaster.
P M Campbell et al.
Insect biochemistry and molecular biology, 28(7), 501-515 (1998-08-27)
Juvenile hormone esterase (JHE) from the prepupal stage of Drosophila melanogaster was purified about 429-fold to near homogeneity by selective precipitations, isoelectric focussing, anion exchange and gel filtration chromatography. The KM and Vmax of the purified enzyme for juvenile hormone
Uppeandra Dhar et al.
Plant cell reports, 24(4), 195-200 (2005-03-12)
A callus induction and in vitro plantlet regeneration system for the endangered state flower of Uttaranchal (Saussurea obvallata) was optimized by studying the influence of explant type (root, hypocotyl, cotyledon and leaf), age and different concentrations of plant growth regulators.
Hye Kyung Kim et al.
Acta crystallographica. Section D, Biological crystallography, 61(Pt 7), 920-931 (2005-06-29)
The enzyme dienelactone hydrolase (DLH) has undergone directed evolution to produce a series of mutant proteins that have enhanced activity towards the non-physiological substrates alpha-naphthyl acetate and p-nitrophenyl acetate. In terms of steady-state kinetics, the mutations caused a drop in
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service