Skip to Content
Merck
All Photos(2)

Key Documents

395B

Sigma-Aldrich

Periodic Acid-Schiff (PAS) Staining System

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352106
NACRES:
NA.47

shelf life

Expiry date on the label.

Quality Level

IVD

for in vitro diagnostic use

application(s)

hematology
histology

shipped in

wet ice

storage temp.

2-8°C

Application

Standard procedure and microwave procedure included in package insert.
When treated with periodic acid, glycols are oxidized to aldehydes. After reaction with Schiff′s Reagent (a mixture of pararosaniline and sodium metabisulfite), a pararosaniline adduct is released that stains the glycol-containing cellular elements pink to red. Cellular elements which may be demonstrated with the PAS procedure include glycogen, fungal walls, basement membrane, certain epithelial sulfomucins and sialomucins, neutral mucosubstances, colloid material of the thyroid and the pars intermedia of the pituitary.

Kit Components Only

Product No.
Description

  • Hematoxylin Solution, Gill No. 3 (kit only) 100 mL

  • Periodic Acid Solution (395-1) 2 x 100

  • Schiff's Reagent (kit only) 4 x 50

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 1B - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1 - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ming Jiang et al.
Nature, 550(7677), 529-533 (2017-10-12)
In several organ systems, the transitional zone between different types of epithelium is a hotspot for pre-neoplastic metaplasia and malignancy, but the cells of origin for these metaplastic epithelia and subsequent malignancies remain unknown. In the case of Barrett's oesophagus
Luke Whitesell et al.
Nature communications, 10(1), 402-402 (2019-01-27)
New strategies are needed to counter the escalating threat posed by drug-resistant fungi. The molecular chaperone Hsp90 affords a promising target because it supports survival, virulence and drug-resistance across diverse pathogens. Inhibitors of human Hsp90 under development as anticancer therapeutics
Satish Patel et al.
PloS one, 6(1), e15845-e15845 (2011-01-22)
Over-activity and elevated expression of glycogen synthase kinase-3 (GSK-3) has been implicated in the etiology of insulin resistance and Type 2 diabetes. Administration of specific GSK-3 inhibitors to diabetic or obese rodent models improves glycaemic control and insulin sensitivity. However
Tavia Caplan et al.
Cell chemical biology, 27(3), 269-282 (2020-01-12)
New strategies are urgently needed to counter the threat to human health posed by drug-resistant fungi. To explore an as-yet unexploited target space for antifungals, we screened a library of protein kinase inhibitors for the ability to reverse resistance of
Anja Beckers et al.
Development (Cambridge, England), 147(21) (2020-05-08)
Cilia are complex cellular protrusions consisting of hundreds of proteins. Defects in ciliary structure and function, many of which have not been characterised molecularly, cause ciliopathies: a heterogeneous group of human syndromes. Here, we report on the FOXJ1 target gene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service