Skip to Content
Merck
All Photos(1)

Key Documents

905

Sigma-Aldrich

Naphthol AS-D Chloroacetate

Synonym(s):

Naphthol AS-D chloroacetate, 3-Hydroxy-2-naphthoic-o-toluidide chloroacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H16ClNO3
CAS Number:
Molecular Weight:
353.80
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.47

form

powder

Quality Level

IVD

for in vitro diagnostic use

application(s)

hematology
histology

shipped in

dry ice

storage temp.

−20°C

SMILES string

Cc1ccccc1NC(=O)c2cc3ccccc3cc2OC(=O)CCl

InChI

1S/C20H16ClNO3/c1-13-6-2-5-9-17(13)22-20(24)16-10-14-7-3-4-8-15(14)11-18(16)25-19(23)12-21/h2-11H,12H2,1H3,(H,22,24)

InChI key

FMVKYSCWHDVMGO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Components

Gelatin capsules contain 20 mg

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M R Baer et al.
Cancer, 63(11), 2192-2200 (1989-06-01)
Leukemia cutis was documented by biopsy in 18 of 877 patients (2%) with acute myelogenous leukemia (AML) seen at Roswell Park Memorial Institute (Buffalo, NY) between 1969 and 1986. French-American-British (FAB) types included four M2, one M3, ten M4, and
Usefulness of naphthol-AS-D-chloroacetate esterase (NASDCAE) in the diagnosis of AML: report of a case.
D Mastrangelo et al.
Haematologica, 70(5), 428-430 (1985-09-01)
Ruliang Xu et al.
Archives of pathology & laboratory medicine, 126(5), 562-566 (2002-04-18)
Placental transmogrification of the lung is a term introduced to describe a peculiar histologic pattern characterized by formation of placental villuslike structures in the lung parenchyma. It has been reported to occur in association with bullous emphysema and lipomatosis. To
K W Lam et al.
Stain technology, 60(3), 169-172 (1985-05-01)
A new cytochemical method is described for the simultaneous demonstration of nonspecific esterase in monocytes and chloracetate esterase in granulocytes. The procedure uses both alpha-naphthyl butyrate and naphthol AS-D chloroacetate as substrates and hexazotized pararosaniline as the coupler. The enzyme
S Jolly et al.
Journal of comparative pathology, 122(2-3), 155-162 (2000-02-24)
The presence of chymase-like proteinase in bovine mast cells was investigated by an enzyme-histochemical technique (naphthol-AS-D-chloroacetate as substrate) in normal skin, primary bronchus, lung and duodenum. The counts and distribution of chymase-positive and toluidine blue-positive mast cells were compared by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service