Skip to Content
Merck
All Photos(1)

Key Documents

08172

Sigma-Aldrich

Cycloartenol

≥90% (GC)

Synonym(s):

(3S,5R,8S,9S,10R,13R,14S,17R)-17-((R)-1,5-Dimethyl-hex-4-enyl)-4,4,13,14-tetramethyl-tetradecahydro-cyclopropa[9,10]cyclopenta[a]phenanthren-3-ol, 9,19-Cyclo-24-lanosten-3β-ol, Handianol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H50O
CAS Number:
Molecular Weight:
426.72
Beilstein:
2224850
UNSPSC Code:
12352211
NACRES:
NA.77

Quality Level

Assay

≥90% (GC)

form

powder

SMILES string

O[C@@H]1C([C@H]2[C@@]3([C@]4([C@H]([C@]5([C@@]([C@H](CC5)[C@@H](CCC=C(C)C)C)(CC4)C)C)CC2)C3)CC1)(C)C

InChI

1S/C30H50O/c1-20(2)9-8-10-21(3)22-13-15-28(7)24-12-11-23-26(4,5)25(31)14-16-29(23)19-30(24,29)18-17-27(22,28)6/h9,21-25,31H,8,10-19H2,1-7H3/t21-,22-,23+,24+,25+,27-,28+,29-,30+/m1/s1

InChI key

ONQRKEUAIJMULO-YBXTVTTCSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

  • Genome-Wide Investigation of Oxidosqualene Cyclase Genes Deciphers the Genetic Basis of Triterpene Biosynthesis in Tea Plants - Research on cycloartenol′s synthesis pathways through the genetic study of oxidosqualene cyclase in tea plants, providing insights into the enhancement of plant sterols beneficial for human health (Du et al., 2024).

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kiyoshi Ohyama et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(3), 725-730 (2009-01-14)
The differences between the biosynthesis of sterols in higher plants and yeast/mammals are believed to originate at the cyclization step of oxidosqualene, which is cyclized to cycloartenol in higher plants and lanosterol in yeast/mammals. Recently, lanosterol synthase genes were identified
Anita Loeschcke et al.
PloS one, 12(12), e0189816-e0189816 (2017-12-28)
Cyclic triterpenes constitute one of the most diverse groups of plant natural products. Besides the intriguing biochemistry of their biosynthetic pathways, plant triterpenes exhibit versatile bioactivities, including antimicrobial effects against plant and human pathogens. While prokaryotes have been extensively used
X Qi et al.
Proceedings of the National Academy of Sciences of the United States of America, 101(21), 8233-8238 (2004-05-19)
The evolution of the ability to synthesize specialized metabolites is likely to have been key for survival and diversification of different plant species. Oats (Avena spp.) produce antimicrobial triterpenoids (avenacins) that protect against disease. The oat beta-amyrin synthase gene AsbAS1
Laurent F Wentzinger et al.
Plant physiology, 130(1), 334-346 (2002-09-13)
To get some insight into the regulatory mechanisms controlling the sterol branch of the mevalonate pathway, tobacco (Nicotiana tabacum cv Bright Yellow-2) cell suspensions were treated with squalestatin-1 and terbinafine, two specific inhibitors of squalene synthase (SQS) and squalene epoxidase
Ryousuke Ito et al.
Organic letters, 13(10), 2678-2681 (2011-04-30)
The gene products of AK121211, AK066327, and AK070534 from Oryza sativa encode cycloartenol, parkeol, and achilleol B synthases, respectively. Parkeol synthase is a unique enzyme that affords parkeol as a single product. Achilleol B synthase is the third seco-type triterpene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service