Skip to Content
Merck
All Photos(1)

Key Documents

D6513

Sigma-Aldrich

Desmosterol

≥84% (GC)

Synonym(s):

24-Dehydrocholesterol, 3β-Hydroxy-5,24-cholestadiene, 5,24-Cholestadien-3β-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H44O
CAS Number:
Molecular Weight:
384.64
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥84% (GC)

form

powder

storage temp.

−20°C

SMILES string

[H][C@@]12[C@]([C@](CC[C@H](O)C3)(C)C3=CC2)([H])CC[C@@]4(C)[C@@]1([H])CC[C@]4([H])[C@]([H])(C)CCC=C(C)C

InChI

1S/C27H44O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h7,9,19,21-25,28H,6,8,10-17H2,1-5H3/t19-,21+,22+,23-,24+,25+,26+,27-/m1/s1

InChI key

AVSXSVCZWQODGV-DPAQBDIFSA-N

Related Categories

General description

Desmosterol is a C27 sterol intermediate in cholesterol synthesis. It may be associated with the pathogenesis of Alzheimer′s disease (AD).[1]

Application

Desmosterol has been used as a standard for the estimation of the concentrations of cholesterol and plant sterols.[2][3] It has also been used to suppress nigericin-induced sterol regulatory element-binding proteins 2 (SREBP2) maturation.[4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anti-atherogenic effects of phytosteryl oleates in apo-E deficient mice
Moghadasian MH, et al
Journal of functional foods, 21, 97-103 (2016)
Clare L Beasley et al.
Bipolar disorders, 7(5), 449-455 (2005-09-24)
Cholesterol forms an integral part of cell membranes and is a major component of myelin. Furthermore, cholesterol also plays a vital role in the development, function and stability of synapses. While low serum cholesterol has previously been associated with mood
Anna Meljon et al.
Journal of lipid research, 53(11), 2469-2483 (2012-08-15)
Unesterified cholesterol is a major component of plasma membranes. In the brain of the adult, it is mostly found in myelin sheaths, where it plays a major architectural role. In the newborn mouse, little myelination of neurons has occurred, and
Ana Rita Lima et al.
Metabolites, 8(1) (2018-03-23)
Prostate cancer (PCa) is an important health problem worldwide. Diagnosis and management of PCa is very complex because the detection of serum prostate specific antigen (PSA) has several drawbacks. Metabolomics brings promise for cancer biomarker discovery and for better understanding
Tim Vanmierlo et al.
Plant foods for human nutrition (Dordrecht, Netherlands), 66(2), 149-156 (2011-03-25)
Plant sterols such as sitosterol and campesterol are frequently applied as functional food in the prevention of atherosclerosis. Recently, it became clear that plasma derived plant sterols accumulate in murine brains. We questioned whether plant sterols in the brain are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service