Skip to Content
Merck
All Photos(4)

Documents

S1270

Sigma-Aldrich

β-Sitosterol

synthetic, ≥95%

Synonym(s):

beta-Sitosterol, α-Dihydrofucosterol, 22,23-Dihydrostigmasterol, 24α-Ethylcholesterol, 5-Stigmasten-3β-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H50O
CAS Number:
Molecular Weight:
414.71
Beilstein:
1916165
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic

Assay

≥95%

form

powder

mp

136-140 °C (lit.)

functional group

hydroxyl

shipped in

ambient

storage temp.

−20°C

SMILES string

O[C@H](C1)CC[C@@]2(C)C1=CC[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@]4([H])[C@H](C)CC[C@H](C(C)C)CC

InChI

1S/C29H50O/c1-7-21(19(2)3)9-8-20(4)25-12-13-26-24-11-10-22-18-23(30)14-16-28(22,5)27(24)15-17-29(25,26)6/h10,19-21,23-27,30H,7-9,11-18H2,1-6H3/t20-,21-,23+,24+,25-,26+,27+,28+,29-/m1/s1

InChI key

KZJWDPNRJALLNS-VJSFXXLFSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

β-Sitosterol has been used:
  • to study its ability to improve the suitability of two pollens
  • as a standard in the determination of sterol purities
  • as a standard in quantitative genetic analysis
  • as a standard in the estimation of plant sterols

Biochem/physiol Actions

A phytosterol with structure very similar to cholesterol that exhibits estrogenic activity. Inhibits proliferation of human leukemia cells, with G2/M arrest, endoreduplication, and polymerization of α-tubulin and microtubules.
Increased β-sitosterol administration displaces cholesterol during absorption. This elevates fecal secretion. It is known to regulate intestinal immunity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Essence of Anesthesia Practice E-Book, 657-657 (2010)
Dong-Oh Moon et al.
Cancer letters, 264(2), 181-191 (2008-03-04)
beta-Sitosterol (SITO) is a potentially valuable candidate for cancer chemotherapy, however the cellular and molecular mechanisms responsible for its anti-cancer activity are unknown. Therefore, we attempted to elucidate the mechanisms responsible for SITO-induced anti-proliferation in human leukemia cells. Treatment with
Sterol limitation in a pollen-fed omnivorous lady beetle (Coleoptera: Coccinellidae)
Pilorget L, et al.
Journal of Insect Physiology, 56(1), 81-87 (2010)
Lipid Analyses of Four Types of Fish From Manitoba Lakes
Moghadasian MH, et al.
EC nutrition, 1(2), 41-48 (2009)
Polymeric Biomaterials: Medicinal and Pharmaceutical Applications, 2, 162-162 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service