Skip to Content
Merck
All Photos(1)

Key Documents

41719

Sigma-Aldrich

(3S)-2,3-Oxidosqualene

≥97.5% (HPLC)

Synonym(s):

(S)-2,3-Epoxy-2,3-dihydrosqualene, (S)-22,23-Epoxy-2,6,10,15,19,23-hexamethyl-2,6,10,14,18-tetracosapentaene, (S)-Squalene 2,3-epoxide, (S)-Squalene 2,3-oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H50O
CAS Number:
Molecular Weight:
426.72
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.28

Assay

≥97.5% (HPLC)

form

liquid

optical purity

enantiomeric excess: ≥90.0%

suitability

conforms to structure for Proton NMR spectrum

storage temp.

−20°C

SMILES string

CC1(C)O[C@H]1CC/C(C)=C/CC/C(C)=C/CC/C=C(C)/CC/C=C(C)/CCC=C(C)C

InChI

1S/C30H50O/c1-24(2)14-11-17-27(5)20-12-18-25(3)15-9-10-16-26(4)19-13-21-28(6)22-23-29-30(7,8)31-29/h14-16,20-21,29H,9-13,17-19,22-23H2,1-8H3/b25-15+,26-16+,27-20+,28-21+/t29-/m0/s1

InChI key

QYIMSPSDBYKPPY-RSKUXYSASA-N

Related Categories

Application


  • Schizophrenic behavior of 2,3-Oxidosqualene Sterol Cyclase from pig liver towards 2,3-oxidosqualene analogues. Alain Krief and colleagues discuss the variable catalytic behaviors of oxidosqualene cyclase when interacting with different oxidosqualene analogues, suggesting implications for sterol production efficiency and specificity (Krief et al., 2021).


Biochem/physiol Actions

(S)-2,3-Epoxysqualene is an intermediate of many metabolic pathways such as the steroid biosynthesis, sesquiterpenoid and triterpenoid biosynthesis, biosynthesis of plant secondary metabolites, biosynthesis of terpenoids and steroids, biosynthesis of plant hormones, biosynthesis of secondary metabolites, and biosynthesis of antibiotics.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Katherine A Mattos et al.
Cellular microbiology, 16(6), 797-815 (2014-02-21)
We recently showed that Mycobacterium leprae (ML) is able to induce lipid droplet formation in infected macrophages. We herein confirm that cholesterol (Cho) is one of the host lipid molecules that accumulate in ML-infected macrophages and investigate the effects of
Xudong Han et al.
Frontiers in oncology, 10, 1549-1549 (2020-10-20)
Glioblastoma, also known as glioblastoma multiforme (GBM), is the most malignant form of glioma and represents 81% of malignant brain and central nervous system (CNS) tumors. Like most cancers, GBM causes metabolic recombination to promote cell survival, proliferation, and invasion
Z Shi et al.
Proceedings of the National Academy of Sciences of the United States of America, 91(15), 7370-7374 (1994-07-19)
The ERG7 gene encoding oxidosqualene-lanosterol cyclase [(S)-2,3-epoxysqualene mutase (cyclizing, lanosterol forming), EC 5.4.99.7] from Saccharomyces cerevisiae has been cloned by genetic complementation of a cyclase-deficient erg7 strain. The DNA sequence of this gene has been determined and found to contain
Deletion of the Gly600 residue of Alicyclobacillus acidocaldarius squalene cyclase alters the substrate specificity into that of the eukaryotic-type cyclase specific to (3S)-2,3-oxidosqualene.
Tsutomu Hoshino et al.
Angewandte Chemie (International ed. in English), 43(48), 6700-6703 (2004-12-14)
Enzymatic cyclization of squalene and oxidosqualene to sterols and triterpenes.
Abe, I., et al.
Chemical Reviews, 93, 2189-2206 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service