Skip to Content
Merck
All Photos(1)

Key Documents

00898

Sigma-Aldrich

DEAE-Dextran

average mol wt 10,000

Synonym(s):

Diethylaminoethyl-Dextran

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12171500
NACRES:
NA.32

form

powder

Quality Level

mol wt

average mol wt 10,000

composition

nitrogen content, 2.5-4.5%

loss

≤10% loss on drying

storage temp.

−20°C

InChI

1S/C11H14O2/c1-3-8-13-11(12)10-6-4-9(2)5-7-10/h4-7H,3,8H2,1-2H3

InChI key

LRNBWFXCDULFQF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service