Skip to Content
Merck
All Photos(1)

Key Documents

93556

Supelco

DEAE-Dextran

average mol wt 500,000

Synonym(s):

Diethylaminoethyl-Dextran

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12171500
NACRES:
NA.32

form

powder

Quality Level

mol wt

average mol wt 500,000

composition

nitrogen content, 2.5-4.5%

loss

≤10% loss on drying

storage temp.

−20°C

InChI

1S/C11H14O2/c1-3-8-13-11(12)10-6-4-9(2)5-7-10/h4-7H,3,8H2,1-2H3

InChI key

LRNBWFXCDULFQF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

DEAE-Dextran is used as mediator in DNA transfection procedures. It is considered to be simple and effective.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthetic DNA delivery systems.
Luo D and Saltzman W M
Nature Biotechnology, 18(1), 33-33 (2000)
Lipofection: a highly efficient, lipid-mediated DNA-transfection procedure.
Felgner P L, et al.
Proceedings of the National Academy of Sciences of the USA, 84(21), 7413-7417 (1987)
Joanne Marie M Del Rosario et al.
Frontiers in immunology, 11, 627-627 (2020-06-18)
Cross-subtype neutralizing single domain antibodies against influenza present new opportunities for immunoprophylaxis and pandemic preparedness. Their simple modular structure and single open reading frame format are highly amenable to gene therapy-mediated delivery. We have previously described R1a-B6, an alpaca-derived single

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service