Skip to Content
Merck
All Photos(1)

Key Documents

90008

Sigma-Aldrich

Blue Dextran

average mol wt 5,000

Synonym(s):

Blue Dextran 5, Dextran blue from Leuconostoc mesenteroides

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.32

biological source

synthetic

form

powder

mol wt

average mol wt 5,000

loss

≤10% loss on drying

capacity

0.01-0.08 mmol/g loading

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service