Skip to Content
Merck
All Photos(3)

Key Documents

Z0625

Sigma-Aldrich

Zinc acetate dihydrate

reagent grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Zn(CH3COO)2 · 2H2O
CAS Number:
Molecular Weight:
219.51
Beilstein:
3732513
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥98% (EDTA titration)
grade:
reagent grade
form:
powder

grade

reagent grade

Quality Level

Assay

≥98% (EDTA titration)

form

powder

reaction suitability

reagent type: catalyst
core: zinc

SMILES string

O.O.CC(=O)O[Zn]OC(C)=O

InChI

1S/2C2H4O2.2H2O.Zn/c2*1-2(3)4;;;/h2*1H3,(H,3,4);2*1H2;/q;;;;+2/p-2

InChI key

BEAZKUGSCHFXIQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

  • Innovative surfactant-free synthesis of core-shell SiO(2)/ZnO particles: rapid ultrasonication and photocatalytic inhibition.: This study utilizes zinc acetate dihydrate in the surfactant-free synthesis of SiO2/ZnO core-shell particles, demonstrating significant photocatalytic properties for environmental applications. The method involves rapid ultrasonication, highlighting the efficiency of zinc acetate dihydrate in advanced material synthesis (Qomariyah et al., 2024).
  • Self-Healing Ionogel-Enabled Self-Healing and Wide-Temperature Flexible Zinc-Air Batteries with Ultra-Long Cycling Lives.: Zinc acetate dihydrate is employed in the fabrication of innovative self-healing ionogels for zinc-air batteries. This development extends battery life and enhances performance across a wide temperature range, marking a breakthrough in energy storage technology (Li et al., 2024).
  • The implementation of ZnS-SnS BM NPs for phenanthrene degradation: An adsorptive photocatalyst approach and its toxicity studies in adult zebrafish.: Zinc acetate dihydrate is a key precursor in synthesizing ZnS-SnS nanoparticles, used for degrading environmental pollutants like phenanthrene. The study also evaluates the ecological impact, demonstrating the material′s low toxicity (K et al., 2024).
  • Effect of ZnCl2 assisted chemical bath deposition on preferred orientations and optical properties of ZnO films.: This research investigates the role of zinc acetate dihydrate in the chemical bath deposition of ZnO films, enhancing their optical properties. The study contributes to the advancement of materials for optical and electronic applications (Guo et al., 2023).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alim Solmaz et al.
Langmuir : the ACS journal of surfaces and colloids, 28(28), 10592-10596 (2012-06-15)
Tapping mode atomic force microscopy (TM-AFM) enables mapping of chemical composition at the nanoscale by taking advantage of the variation in phase angle shift arising from an embedded second phase. We demonstrate that phase contrast can be attributed to the
Pier Francesco Porciani et al.
The Journal of clinical dentistry, 23(3), 76-79 (2012-12-06)
A controlled, clinical, double-blind study was conducted to assess the efficacy of a sugar-free chewing gum containing zinc acetate and magnolia bark extract (MBE) on oral volatile sulfur-containing compounds (VSC) versus a placebo sugar-free chewing gum for two hours. To
Robert Krysiak et al.
The Yale journal of biology and medicine, 85(2), 249-254 (2012-06-28)
Wilson's disease is a rare genetic disorder of copper metabolism. The difference in copper tissue accumulation is responsible for the various clinical manifestations of this disorder. If left untreated, Wilson's disease progresses to hepatic failure, severe neurological disability, and even
Yoshiko Uesugi et al.
Journal of drug targeting, 20(3), 224-234 (2011-11-18)
This study is undertaken to design zinc-stabilized gelatin nano-complexes of tissue-type plasminogen activator (t-PA) for thrombolytic therapy where the t-PA activity can be recovered in the blood circulation upon ultrasound irradiation. Various molecular weights of gelatin were complexed with t-PA
R Dhariwal et al.
Journal of the Indian Society of Pedodontics and Preventive Dentistry, 30(1), 85-88 (2012-05-09)
Oral submucous fibrosis is a chronic debilitating disorder which is usually seen in adults with areca nut chewing habit. A rapid upsurge in the popularity of commercial areca nut products, especially among the younger generation, is a cause of grave

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service