Skip to Content
Merck
All Photos(2)

Key Documents

379786

Sigma-Aldrich

Zinc acetate dihydrate

99.999% trace metals basis

Synonym(s):

Dicarbomethoxyzinc, Zinc diacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Zn(CH3COO)2 · 2H2O
CAS Number:
Molecular Weight:
219.51
Beilstein:
3732513
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99.999% trace metals basis

form

powder or chunks

SMILES string

O.O.CC(=O)O[Zn]OC(C)=O

InChI

1S/2C2H4O2.2H2O.Zn/c2*1-2(3)4;;;/h2*1H3,(H,3,4);2*1H2;/q;;;;+2/p-2

InChI key

BEAZKUGSCHFXIQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Zinc acetate dihydrate is widely used as a precursor to synthesize ZnO nanomaterials. For example, high-purity single crystal ZnO nanowires can be prepared by thermal decomposition of zinc acetate dihydrate at 300 0C in air.

It can be used to prepare solution-processable ZnO semiconductor ink and an inverted staggered type thin film transistor can be fabricated by spin-casting ZnO ink on a Si wafer.

It can also be used to enhance the structural and optical properties of TiO2–ZnO core-shell nanograss by controlling their size and morphology.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Crystal Structure of Zinc Acetate Dihydrate~ Zn(CH3COO)2.2 H20
Van Niekerk JN, et al.
Acta Crystallographica, 6 (1953)
Effect of zinc acetate dihydrate precursor concentration on the properties of TiO2-ZnO core-shell nanograss hetero-structure.
Samsuri SAM, et al.
J. Alloy Compounds, 623, 460-465 (2015)
Zhang, B. et al.
Inorganic Chemistry, 37, 1844-1844 (1998)
Dhas, N.A. et al.
Chemistry of Materials, 11, 806-806 (1999)
Pier Francesco Porciani et al.
The Journal of clinical dentistry, 23(3), 76-79 (2012-12-06)
A controlled, clinical, double-blind study was conducted to assess the efficacy of a sugar-free chewing gum containing zinc acetate and magnolia bark extract (MBE) on oral volatile sulfur-containing compounds (VSC) versus a placebo sugar-free chewing gum for two hours. To

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service