Skip to Content
Merck
All Photos(1)

Key Documents

32526

Supelco

Eprinomectin

PESTANAL®, analytical standard, mixture of isomers

Synonym(s):

(4′’R)-4″-(Acetylamino)-4″-deoxy-avermectin B1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C50H75NO14
CAS Number:
Molecular Weight:
914.13
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

quality

mixture of isomers

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

−20°C

SMILES string

C[C@H]1[C@H](NC(C)=O)[C@@H](OC)C[C@H](O[C@H]2[C@H](C)O[C@@H](O[C@H](/C(C)=C/C[C@]3([H])C[C@H]4C[C@@]5(O[C@@]([R])([H])[C@@H](C)C=C5)O3)[C@@H](C)/C=C/C=C(CO6)/[C@@]([C@@]6([H])[C@H](O)C(C)=C7)(O)[C@]7([H])C(O4)=O)C[C@@H]2OC)O1.CCC.CCCC

InChI

1S/C49H73NO14/c1-25(2)43-28(5)17-18-48(64-43)23-35-20-34(63-48)16-15-27(4)44(26(3)13-12-14-33-24-57-46-42(52)29(6)19-36(47(53)60-35)49(33,46)54)61-40-22-38(56-11)45(31(8)59-40)62-39-21-37(55-10)41(30(7)58-39)50-32(9)51/h12-15,17-19,25-26,28,30-31,34-46,52,54H,16,20-24H2,1-11H3,(H,50,51)/b13-12-,27-15-,33-14-/t26-,28-,30-,31-,34+,35-,36-,37+,38-,39-,40-,41+,42+,43+,44-,45+,46+,48+,49+/m0/s1

InChI key

WPNHOHPRXXCPRA-TVXIRPTOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Eprinomectin is a second generation macrocyclic lactone, used as a topical endectocide which is active both against endoparasites and ectoparasites for all cattle, including lactating dairy cows.

Application

Eprinomectin has been used as a standard for the determination of pesticide residues in soil samples by the on-line solid-phase extraction-ultra high performance liquid chromatography-tandem mass spectrometry. It may also be used to examine the presence of any veterinary drug residues in the bulk milk tank.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eprinomectin: a novel avermectin for use as a topical endectocide for cattle.
Shoop W L, et al.
International Journal For Parasitology, 26(11), 1237-1242 (1996)
W A Mason et al.
Veterinary parasitology, 189(2-4), 250-259 (2012-06-06)
A randomised clinical trial from the North Island of New Zealand was conducted to assess the effect of repeated anthelmintic treatment on milk production, and to assess factors that affect treatment response. Nine hundred and twenty three multiparous, lactating dairy
Huiqiang Wen et al.
Veterinary parasitology, 174(1-2), 72-76 (2010-09-21)
Chinese Holstein, bred by mating the Holstein-Friesian to Chinese Yellow Cattle, is a major dairy cattle breed in China. Eprinomectin is widely used in the treatment of nematode and ectoparasite infections in lactating cattle. The pharmacokinetics of eprinomectin in the
Solange Kiki-Mvouaka et al.
Drug metabolism and disposition: the biological fate of chemicals, 38(4), 573-580 (2010-01-22)
Macrocyclic lactones (MLs) are lipophilic anthelmintics and substrates for P-glycoprotein (P-gp), an ATP-binding cassette transporter involved in drug efflux out of both host and parasites. To evaluate the contribution of P-gp to the in vivo kinetic disposition of MLs, the
M D Soll et al.
Veterinary parasitology, 192(4), 313-320 (2013-01-01)
A series of 10 dose confirmation studies was conducted to evaluate the persistent activity of an extended-release injectable (ERI) formulation of eprinomectin against single point challenge infections of gastrointestinal and pulmonary nematodes of cattle. The formulation, selected based on the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service