Skip to Content
Merck
All Photos(3)

Key Documents

228737

Sigma-Aldrich

Zinc nitrate hexahydrate

reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Zn(NO3)2 · 6H2O
CAS Number:
Molecular Weight:
297.49
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
98%
grade:
reagent grade
form:
flakes
powder or crystals
solubility:
water: soluble(lit.)
Pricing and availability is not currently available.

grade

reagent grade

Quality Level

Assay

98%

form

flakes
powder or crystals

solubility

water: soluble(lit.)

SMILES string

O.O.O.O.O.O.[Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/2NO3.6H2O.Zn/c2*2-1(3)4;;;;;;;/h;;6*1H2;/q2*-1;;;;;;;+2

InChI key

JGPSMWXKRPZZRG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used to synthesize a new microporous, organically templated zinc phosphate. Such compounds have potential applications as catalysts or in other applications involving microporosity.[1]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toward wearable and stretchable fabric-based supercapacitors: novel ZnO and SnO2 nanowires-carbon fibre and carbon paper hybrid structure.
Bae J, et al.
Journal of Solid State Electrochemistry, 19(1), 211-219 (2015)
Harmon, S.B. Sevov, S.C.
Chemistry of Materials, 10, 3020-3020 (1998)
Robert J Friederichs et al.
Journal of the Royal Society, Interface, 12(108), 20150190-20150190 (2015-06-05)
Experimental chemistry and atomic modelling studies were performed here to investigate a novel ionic co-substitution in hydroxyapatite (HA). Zinc, silicate co-substituted HA (ZnSiHA) remained phase pure after heating to 1100 °C with Zn and Si amounts of 0.6 wt% and
Milna GWA.
Gardner's Commercially Important Chemicals: Synonyms, Trade Names, and Properties, 4153-4153 (2015)
Structural and DC electrical investigations of ZnO thin films prepared by spray pyrolysis technique.
Riad AS, et al.
Physica B: Condensed Matter (Amsterdam, Netherlands), 296(4), 319-325 (2001)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service