Skip to Content
Merck
All Photos(1)

Key Documents

650498

Sigma-Aldrich

Chloroform

HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer

Synonym(s):

Methylidyne trichloride, Trichloromethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CHCl3
CAS Number:
Molecular Weight:
119.38
Beilstein:
1731042
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.04
grade:
HPLC Plus
for residue analysis
technique(s):
HPLC: suitable
gas chromatography (GC): suitable
application(s):
food and beverages
bp:
60.5-61.5 °C (lit.)
vapor pressure:
160 mmHg ( 20 °C)

grade

HPLC Plus
for residue analysis

vapor density

4.1 (vs air)

vapor pressure

160 mmHg ( 20 °C)

Assay

≥99.9%

form

liquid

contains

amylenes as stabilizer

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.02% water
≤0.5 ppb Fluorescence (quinine) at 365 nm

evapn. residue

≤0.0001%

halogenated residue

<10 ng/L (as heptachlor epoxide)

refractive index

n20/D 1.445 (lit.)

bp

60.5-61.5 °C (lit.)

mp

−63 °C (lit.)

density

1.48 g/mL at 25 °C
1.492 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 245 nm Amax: 1.00
λ: 250 nm Amax: 0.30
λ: 275-400 nm Amax: 0.005

suitability

suitable for residue analysis

application(s)

food and beverages

SMILES string

ClC(Cl)Cl

InChI

1S/CHCl3/c2-1(3)4/h1H

InChI key

HEDRZPFGACZZDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chloroform is an organic chlorinated hydrocarbon (CHC) commonly used in industries as a solvent.

Application

Chloroform may be used in the following:

  • Extraction of active constituents of anti-inflammatory drugs for their analysis by attenuated total reflectance-Fourier transform infrared spectroscopic (ATR-FTIR) methods
  • Estimation of ascorbic acid, free phenolic acids, and flavonoids in extracts of chloroform, methanol, water and, acetic acid obtained from two edible leaf samples using reversed-phase high-performance liquid chromatography (RP-HPLC) coupled to photodiode array detector (DAD)
  • Preparation of cannabis product extracts for the analysis of delta-9-tetrahydrocannabinol (THC) residues on hands of suspected consumers by ion-mobility spectrometry (IMS)
  • Investigation of different solvents to measure their recovery ratios for the extraction of acrylamide and 5-hydroxymethylfurfural (HMF) from seafood samples to develop an HPLC-MS/MS-based method for their quantification
  • Extraction of pesticides by direct-immersion single drop micro-extraction (SDME) method from mango samples for their subsequent analysis by GC-MS
  • Sample preparation in the profiling of lipids from complex biological samples using off-line mixed-mode liquid chromatography coupled with reversed phase high performance liquid chromatography-high resolution mass spectrometry

Chloroform was used in the making of silanized coverslips for DNA combing.
It may be used as:
  • Extractant in solvent extraction process.
  • A component of the binary mobile phases for liquid chromatography–atmospheric pressure photoionization-mass spectrometry (LC–APPIMS) of triacylglycerol model compounds.
  • Solvent for recrystallization.
Suitable for HPLC, spectrophotometry, environmental testing

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silanization of coverslips for DNA combing.
Labit H, et al.
Biotechniques, 11, 50-50 (2009)
Sheng-Suan Cai et al.
Journal of chromatography. A, 1173(1-2), 88-97 (2007-10-30)
In this work, we evaluate the performance of liquid chromatography-atmospheric pressure photoionization-mass spectrometry (LC-APPI-MS) for non-aqueous reversed phase analysis of six triacylglycerol model compounds using six binary mobile phases including MeOH/iPrOH, MeOH/CHCl(3), MeOH/CH(2)Cl(2), CH(3)CN/iPrOH, CH(3)CN/CHCl(3), and CH(3)CN/CH(2)Cl(2). All mobile phases
Electrogenerated chemiluminescent emission from an organized (L-B) monolayer of a Ru(bpy)32+-based surfactant on semiconductor and metal electrodes.
Zhang X and Bard AJ.
The Journal of Physical Chemistry, 92(20), 5566-5569 (1988)
Analytical chemistry in a drop. Solvent extraction in a microdrop.
Liu H and Dasgupta PK.
Analytical Chemistry, 68(11), 1817-1821 (1996)
Kuan Sun et al.
Nature communications, 6, 6013-6013 (2015-01-15)
Solution-processed organic photovoltaic cells (OPVs) hold great promise to enable roll-to-roll printing of environmentally friendly, mechanically flexible and cost-effective photovoltaic devices. Nevertheless, many high-performing systems show best power conversion efficiencies (PCEs) with a thin active layer (thickness is ~100 nm) that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service