Skip to Content
Merck
All Photos(1)

Documents

C2284

Sigma-Aldrich

Copper(II) sulfate solution

4 % (w/v) (prepared from copper (II) sulfate pentahydrate)

Synonym(s):

Cupric sulfate standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55

form

liquid

reaction suitability

reagent type: catalyst
core: copper

concentration

4 % (w/v) (prepared from copper (II) sulfate pentahydrate)

pH

4

suitability

suitable for determination of total protein (Recommended use with BCA solution (B9643) for total protein determination.)

storage temp.

room temp

SMILES string

[Cu++].[O-]S([O-])(=O)=O

InChI

1S/Cu.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

ARUVKPQLZAKDPS-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application


  • Structural Simulation and Performance Evaluation of a Novel Synthesized ZIF in the Adsorption of MO from Aqueous Solutions: This paper evaluates a newly synthesized zeolitic imidazolate framework (ZIF), incorporating copper(II) sulfate for enhanced adsorption of methyl orange from water, demonstrating its efficacy in water purification technologies (Ravankhah and Fathi, 2024).

Packaging

Also available as part of the Bicinchoninic Acid Kit for Protein Determination (BCA-1).

Suitability

This product is commonly used for protein determination by the Bicinchoninic Acid (BCA) method.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Motoi Kikusato et al.
Poultry science, 100(2), 957-963 (2021-02-02)
Broiler chickens reared under heat stress (HS) conditions have decreased growth performance and show metabolic and immunologic alterations. This study aimed to evaluate the effect of supplementation with a standardized blend of plant-derived isoquinoline alkaloids (IQ) on the growth performance
Sarah J Routledge et al.
Microbial cell factories, 10, 17-17 (2011-03-24)
Pichia pastoris is a widely-used host for recombinant protein production. Initial screening for both suitable clones and optimum culture conditions is typically carried out in multi-well plates. This is followed by up-scaling either to shake-flasks or continuously stirred tank bioreactors.
Chiara Milani et al.
International journal of molecular sciences, 21(10) (2020-05-28)
In northern Italy, biomass burning-derived (BB) particles and diesel exhaust particles (DEP) are considered the most significant contributors to ultrafine particle (UFP) emission. However, a comparison between their impact on different brain regions was not investigated until now. Therefore, male
David Sanz-Rubio et al.
Scientific reports, 8(1), 10306-10306 (2018-07-10)
Exosomes are nano-vesicles present in the circulation that are involved in cell-to-cell communication and regulation of different biological processes. MicroRNAs (miRNAs) are part of their cargo and are potential biomarkers. Methods of exosome isolation and the inter-individual and intra-individual variations
Mary T Galgano et al.
Human pathology, 40(10), 1434-1440 (2009-05-12)
Protein kinase C-related kinases are regulated by phosphatidylinositol-3-kinase and Rho family GTPases. The isoform PRK1 has been characterized in detail in prostate cancer, but not in other carcinomas. We analyzed our prior microarray data for PRK1 gene expression in 175

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service