Recommended Products
form
liquid
suitability
suitable for determination of protein concentration
General description
Protein determination is one of the most common operations performed in biochemical research. Bicinchoninic acid solution is used in the bicinchoninic acid (BCA) assay, which is equivalent to the Lowry procedure. Both rely on the formation of a Cu2+-protein complex under alkaline conditions, followed by reduction of the Cu2+ to Cu1+. The amount of reduction is proportional to the protein present. The BCA assay is more sensitive and applicable than either biuret or Lowry procedures.
Application
Bicinchoninic Acid solution (Bicinchoninic acid protein assay kit) is used :
- For the determination of total protein content in the process of quantifying bromelain, extracted from the pineapple stem residues using aqueous micellar two-phase systems composed of ionic liquids as co-surfactants.
- For the determination of bovine serum albumin (BSA) in biomimetic calcium phosphate coating co-precipitated on titanium alloy plates.
Features and Benefits
- It is easy to use.
- The color complex is stable.
- There is less susceptibility to detergents.
- It is applicable over a broad range of protein concentrations.
Packaging
Also available as part of the Bicinchoninic Acid Kit for Protein Determination (BCA-1).
Suitability
This product is commonly used for protein determination by the Bicinchoninic Acid (BCA) method.
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Zhenhua Ma et al.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP, 216, 29-37 (2018-11-12)
This study explored the impact of fishmeal replacement by commercial soybean meal (SM) and EnzoMeal (EZM) on Asian seabass Lates calcarifer growth performance using six diets. The six diets comprised two sources of plant proteins with three levels each, including
Shabnam Khatibi et al.
BMC systems biology, 11(1), 48-48 (2017-04-15)
Transforming growth factor β (TGF-β) signalling regulates the development of embryos and tissue homeostasis in adults. In conjunction with other oncogenic changes, long-term perturbation of TGF-β signalling is associated with cancer metastasis. Although TGF-β signalling can be complex, many of
Biomimetic coprecipitation of calcium phosphate and bovine serum albumin on titanium alloy
Liu Y, et al.
Journal of Biomedical Materials Research, 57(3), 327-335 (2001)
Diego García-Ayuso et al.
Experimental eye research, 161, 10-16 (2017-05-30)
In this work we study the effects of an acute light-induced retinal degeneration on the population of melanopsin positive retinal ganglion cells (m
Recovery of bromelain from pineapple stem residues using aqueous micellar two-phase systems with ionic liquids as co-surfactants
Vicente AF, et al.
Process Biochemistry (Oxford, United Kingdom), 51(4), 528-534 (2016)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service