Skip to Content
Merck
All Photos(3)

Key Documents

451657

Sigma-Aldrich

Copper(II) sulfate

anhydrous, powder, ≥99.99% trace metals basis

Synonym(s):

Cupric sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuSO4
CAS Number:
Molecular Weight:
159.61
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
anhydrous
form:
powder

grade

anhydrous

Quality Level

vapor pressure

7.3 mmHg ( 25 °C)

Assay

≥99.99% trace metals basis

form

powder

impurities

≤100.0 ppm Trace Metal Analysis

mp

200 °C (dec.) (lit.)

density

3.603 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

[Cu++].[O-]S([O-])(=O)=O

InChI

1S/Cu.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

ARUVKPQLZAKDPS-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Anhydrous form of copper sulfate occurs as mineral hydrocyanite. It exhibits a rhombic crystalline morphology. It decomposes to its oxide at temperatures above 600oC. Hydrate of CuSO4 is prepared by the addition of dilute sulfuric acid on copper (II) oxide or copper (II) carbonate, blue triclinic crystals of pentahydrate are formed upon crystallization. In industrial preparation of CuSO4, a hot mixture of dilute sulfuric acid and scrap copper is exposed to air flow.

Application

This anhydrous salt may be used in the electroplating and mining industries and to preserve wood in combination chromium and arsenic.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

<SMALL>John Daintith</SMALL> et al.
Chemistry (Weinheim An Der Bergstrasse, Germany) (2005)
<SMALL>Egon Wiberg, Nils Wiberg</SMALL> et al.
Inorganic Chemistry, Academic Press (2001)
<SMALL>H. Wayne Richardson</SMALL> et al.
Handbook of Copper Compounds and Applications null
Lydia Boike et al.
Cell chemical biology, 28(1), 4-13 (2020-09-24)
MYC is a major oncogenic transcriptional driver of most human cancers that has remained intractable to direct targeting because much of MYC is intrinsically disordered. Here, we have performed a cysteine-reactive covalent ligand screen to identify compounds that could disrupt
Hung T Vu et al.
Environmental toxicology and chemistry, 33(4), 791-797 (2013-12-10)
The assumption of the individual effective dose is the basis for the probit method used for analyzing dose or concentration-response data. According to this assumption, each individual has a uniquely innate tolerance expressed as the individual effective dose (IED) or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service