Skip to Content
Merck
All Photos(1)

Documents

BCA1

Sigma-Aldrich

Bicinchoninic Acid Kit for Protein Determination

for 200-1000 μg/ml protein

Synonym(s):

BCA protein assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352106
NACRES:
NA.32

usage

sufficient for ~500 assays

storage temp.

2-8°C

Application

Bicinchoninic Acid Kit has been used for the protein estimation in salivary proteome samples, human embryonic kidney 293 cells and luteal cells.

Features and Benefits

  • Simple, sensitive colorimetric assay for proteins
  • Two stable reagents are used for the working solution
  • Faster and easier than the Lowry protein assay
  • Compatible with many detergents both ionic and nonionic
  • Less variation between different proteins than the Bradford dye-binding assay
  • Adaptable for use in microwell plates

Principle

Proteins reduce alkaline Cu(II) to Cu(I) in a concentration-dependent manner. Bicinchoninic acid is a highly specific chromogenic reagent for Cu(I), forming a purple complex with an absorbance maximum at 562 nm. The absorbance is directly proportional to protein concentration. This is an alternative to the Folin-Ciocalteu reagent for protein determination.

Kit Components Only

Product No.
Description

  • Bicinchoninic Acid Solution 1 L

  • 4%(w/v) CuSO4 · 5H2O Solution 25 mL

  • Protein Standard Solution 5 x 1

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Jantas et al.
Neurotoxicity research, 25(2), 208-225 (2013-10-10)
Tianeptine (Tian) possesses neuroprotective potential, however, little is known about the effect of this drug in models of neuronal apoptosis. In the present study, we aimed (1) to compare the neuroprotective capacities of some antidepressants (ADs) in the models of
TGFB1 modulates in vitro secretory activity and viability of equine luteal cells
Galvao A, et al.
Cytokine, 110, 316-327 (2018)
Salivary proteome patterns of individuals exposed to High Altitude
Jain S, et al.
Archives of Oral Biology, 96, 104-112 (2018)
K J Wiechelman et al.
Analytical biochemistry, 175(1), 231-237 (1988-11-15)
The colored complex formed between Cu+ and bicinchoninic acid is the basis of the bicinchoninic acid protein assay (P. K. Smith, R. I. Krohn, G. T. Hermanson, A. K. Mallia, F. H. Gartner, M. D. Provenzano, E. K. Fujimoto, N.
Human OATP1B1 (SLCO1B1) transports sulfated bile acids and bile salts with particular efficiency
Toth B, et al.
Toxicology in vitro, 52, 189-194 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service