Skip to Content
Merck
All Photos(9)

Key Documents

746479

Sigma-Aldrich

Lithium bromide

anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiBr
CAS Number:
Molecular Weight:
86.85
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
form:
powder

grade

anhydrous

Quality Level

vapor pressure

1 hPa ( 748 °C)

product line

ReagentPlus®
Redi-Dri

Assay

≥99%

form

powder

quality

free-flowing

pH

7 (20 °C, 10 g/L)

mp

550 °C (lit.)

SMILES string

[Li+].[Br-]

InChI

1S/BrH.Li/h1H;/q;+1/p-1

InChI key

AMXOYNBUYSYVKV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Lithium bromide is a versatile reagent that is commonly used as a nucleophilic brominating reagent. It is also employed as a Lewis acid catalyst and a mediator in various reactions.

Application

Lithium bromide can be used as a catalyst in the:
  • Solvent-free condensation of arylene diamines and esters to synthesize 2-substituted benzimidazoles and imidazopyridines.
  • Three-component condensation of aldehydes, β-ketoesters, and urea to synthesize dihydropyrimidinones via Biginelli reaction.
  • Solvent-free Ehrlich-Sachs reaction.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexia F P Lépine et al.
Molecular nutrition & food research, 63(2), e1801007-e1801007 (2018-11-10)
Resistant starches (RSs) are classically considered to elicit health benefits through fermentation. However, it is recently shown that RSs can also support health by direct immune interactions. Therefore, it has been hypothesized that the structural traits of RSs might impact
Mehdi Fathi et al.
Materials science & engineering. C, Materials for biological applications, 103, 109743-109743 (2019-07-28)
Bacterial infection, wide inflammation, and osteoporosis are the most common factors in the failure of orthopedic implants. The present study aims to design an orthopedic implant based on Titania nanotubes (TiO2-NTs) which not only have a high biocompatibility but also
Zhina Hadisi et al.
Macromolecular bioscience, 20(4), e1900328-e1900328 (2020-02-23)
Burn injuries represent a major life-threatening event that impacts the quality of life of patients, and places enormous demands on the global healthcare systems. This study introduces the fabrication and characterization of a novel wound dressing made of core-shell hyaluronic
Majd Al-Naji et al.
ChemSusChem, 12(12), 2628-2636 (2019-04-18)
The need for more sustainable products and processes has led to the use of new methodologies with low carbon footprints. In this work, an efficient tandem process is demonstrated for the liquid-phase catalytic upgrading of lignocellulosic biomass-derived γ-valerolactone (GVL) with
Antonella Motta et al.
Journal of tissue engineering and regenerative medicine, 8(11), 874-885 (2012-09-14)
Silk fibroin fibres from two different sources, Bombyx mori pure-breed silkworms and polyhybrid cross-bred silkworm cocoons, were treated with formic acid under planar stirring conditions to prepare non-woven nets. The treatment partially dissolved the fibres, which bound together and formed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service