Skip to Content
Merck
All Photos(4)

Documents

310468

Sigma-Aldrich

Lithium chloride

ACS reagent, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiCl
CAS Number:
Molecular Weight:
42.39
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Assay

≥99%

form

crystalline powder
powder, crystals or granules

impurities

≤0.008 meq/g Titr. base
≤0.01% insolubles

loss

≤1.0% loss on drying

pH

6

mp

605 °C (lit.)

anion traces

sulfate (SO42-): ≤0.01%

cation traces

Ba: ≤0.003%
Ca: ≤0.01%
Fe: ≤0.001%
K: ≤0.01%
Na: ≤0.20%
heavy metals: ≤0.002% (by ICP)

SMILES string

[Li+].[Cl-]

InChI

1S/ClH.Li/h1H;/q;+1/p-1

InChI key

KWGKDLIKAYFUFQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Lithium chloride (LiCl) is a lithium salt. It is used as a source of chloride nucleophile as well as a source of chloride ligand in chemical synthesis. LiCl is also used as a desiccant in drying air streams.

Application

Lithium chloride can be used as:      
  • A source of chloride anion for the conversion of alcohols to alkyl chloride under Mitsunobu conditions.      
  • An additive in the palladium-catalyzed coupling and carbonylative coupling reactions of organostannanes and vinyl triflates.      
  • A reagent for the cleavage of protecting groups under mild conditions. It readily converts acetals and ketals to the corresponding carbonyl compounds.
  • A reagent in the dehydrohalogenation of α-halocarbonyl compounds to α,β-unsaturated carbonyls compound in the presence of DMF.

LiCl is also used with other reagents:      
  • LiCl:DIPEA (1:1 molar ratio) is used in Horner–Wadsworth–Emmons alkenation reaction.      
  • LiCl:Hexamethylphosphoric triamide is used in decarboxylative alkylation and chloride displacement reactions.
  • LiCl is used as an additive in solid-phase peptide synthesis because it increases resin swelling and improves the efficiencies of complex coupling steps.

Features and Benefits

  • Easy to handle      
  • Weak Lewis acid      
  • Highly soluble in water

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Horner-wadsworth-emmons reaction: Use of lithium chloride and an amine for base-sensitive compounds.
Blanchette MA, et al.
Tetrahedron Letters, 25(21), 2183-2186 (1984)
Derivatization of cellulose in lithium chloride and N-N-dimethylacetamide solutions.
McCormick CL and Callais PA.
Polymer, 28(13), 2317-2323 (1987)
Solution studies of cellulose in lithium chloride and N,N-dimethylacetamide.
McCormick CL, et al.
Macromolecules, 18(12), 2394-2401 (1985)
Hart WA, et al.
The Chemistry of Lithium, Sodium, Potassium, Rubidium, Cesium and Francium, 355-355 (1973)
Eagleson M.
Concise Encyclopedia Chemistry, 605-605 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service