Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lithium bromide
Linear Formula:
LiBr
CAS Number:
7550-35-8
Molecular Weight:
86.85
EC Number:
231-439-8
Products
Product Category
salts (5)
Brand
Sigma-Aldrich (5)
Supelco (1)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Special Grade
Anhydrous (3)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
213225
Lithium bromide,
ReagentPlus
®
, ≥99%
Expand
View Pricing
746479
Lithium bromide,
anhydrous, free-flowing, Redi-Dri
™
,
ReagentPlus
®
, ≥99%
Expand
View Pricing
229733
Lithium bromide,
powder and chunks, ≥99.995% trace metals basis
Expand
View Pricing
429465
Lithium bromide,
AnhydroBeads
™
, −10 mesh, 99.999% trace metals basis
Expand
View Pricing
1.05669
Lithium bromide,
extra pure
Expand
View Pricing
449873
Lithium bromide,
AnhydroBeads
™
, −10 mesh, ≥99.9% trace metals basis
Expand
View Pricing