Skip to Content
Merck
All Photos(3)

Key Documents

229733

Sigma-Aldrich

Lithium bromide

powder and chunks, ≥99.995% trace metals basis

Synonym(s):

Lithium monobromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiBr
CAS Number:
Molecular Weight:
86.85
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
≥99.995% trace metals basis
form:
powder and chunks

Quality Level

Assay

≥99.995% trace metals basis

form

powder and chunks

impurities

≤50.0 ppm Trace Metal Analysis

mp

550 °C (lit.)

application(s)

battery manufacturing

SMILES string

[Li+].[Br-]

InChI

1S/BrH.Li/h1H;/q;+1/p-1

InChI key

AMXOYNBUYSYVKV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Space cooling using geothermal single‐effect water/lithium bromide absorption chiller: This research explores the use of lithium bromide in geothermal absorption chillers for space cooling applications (M El Haj Assad, M Sadeghzadeh, 2021).

  • A facile and fast method for quantitating lignin in lignocellulosic biomass using acidic lithium bromide trihydrate (ALBTH): The paper introduces a novel method for lignin quantification using lithium bromide trihydrate (N Li, X Pan, J Alexander, 2016).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

B Saake et al.
Bioresource technology, 80(3), 195-204 (2001-10-17)
Four xylan samples from different origin were investigated, using a multi-detector, size exclusion, chromatographic system with two chromatographic column sets and mobile phases differing in the DMSO:water ratio. Molar mass distribution could be analysed best using a mobile phase of
Sophie Nocquet-Thibault et al.
Organic letters, 15(8), 1842-1845 (2013-04-02)
Using (diacetoxyiodo)benzene in conjunction with simple bromide salts in ethanol allows the regioselective ethoxybromination of a wide range of enamides, thus yielding highly versatile α-bromo hemiaminals, which can then be engaged in a broad array of transformations.
Mohammad M Mojtahedi et al.
Organic letters, 9(15), 2791-2793 (2007-06-22)
A room temperature convenient disproportionation or reduction of aldehydes prompted by lithium bromide and triethylamine is described in a solvent-free environment. Distribution of the products to selectively direct the process toward Cannizzaro or Tishchenko reactions is controlled by the type
R Quaderer et al.
Organic letters, 3(20), 3181-3184 (2001-09-28)
[reaction: see text] The alkanesulfonamide "safety-catch" resin has proven useful for Fmoc-based synthesis of C-terminal peptide thioesters. We now report that the yield of isolated thioester can increase significantly when the cleavage reaction is carried out in 2 M LiBr/THF
Ngoc-Ly Hoang et al.
Journal of chromatography. A, 1205(1-2), 60-70 (2008-08-30)
The structure of starch molecules is relevant to nutrition and industrial applications. Size-exclusion chromatography (SEC, also known as GPC) of native starch generally suffers non-satisfactory repeatability and reproducibility of the dissolution and separation. This work combines two polar organic solvents:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service