Skip to Content
Merck
All Photos(8)

Key Documents

793574

Sigma-Aldrich

Sodium bromide

anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaBr
CAS Number:
Molecular Weight:
102.89
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
anhydrous
form:
powder

grade

ACS reagent
anhydrous

Quality Level

vapor pressure

1 mmHg ( 806 °C)

product line

Redi-Dri

Assay

≥99%

form

powder

quality

free-flowing

impurities

≤0.005% insolubles

pH

5.0-8.8 (25 °C, 5%)

mp

755 °C (lit.)

anion traces

bromate (BrO3-): ≤0.001%
chloride (Cl-): ≤0.2%
sulfate (SO42-): ≤0.002%

cation traces

Ba: ≤0.002%
Ca: ≤0.002%
Fe: ≤5 ppm
K: ≤0.1%
Mg: ≤0.001%
heavy metals: ≤5 ppm (by ICP-OES)

SMILES string

[Na+].[Br-]

InChI

1S/BrH.Na/h1H;/q;+1/p-1

InChI key

JHJLBTNAGRQEKS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sodium bromide (NaBr) is an inorganic salt. It is an inexpensive source of bromide anion. It is used as a nucleophilic catalyst in organic synthesis. Additionally, NaBr is a useful electrolyte in electrochemical oxidations.

Application


  • Sensitive and Low-Noise Perovskite Nanocrystal-Organic Bulk Heterostructure X-ray Detectors Enabled by Sodium Bromide-Assisted In Situ Reparation: This research underscores the role of sodium bromide in enhancing the fabrication of perovskite nanocrystals for advanced X-ray detection technologies, illustrating its utility in high-precision imaging applications critical for both medical and industrial use (Li Y, Fang Y, 2024).

  • Surface-Modified Carboxylated Cellulose Nanofiber Hydrogels for Prolonged Release of Polyhexamethylene Biguanide Hydrochloride (PHMB) for Antimicrobial Applications: Demonstrates the effectiveness of sodium bromide in modifying cellulose nanofibers for use in sustained drug release systems, highlighting its importance in the development of antimicrobial materials (O-Chongpian P, Jantrawut P, 2023).

  • Unravelling Alkali-Metal-Assisted Domain Distribution of Quasi-2D Perovskites for Cascade Energy Transfer toward Efficient Blue Light-Emitting Diodes: Details the utilization of sodium bromide in the synthesis of quasi-2D perovskites, paving the way for improvements in blue light-emitting diode (LED) technology which is crucial for display and lighting solutions (Cai W, Yip HL, 2022).

  • Chronic sodium bromide treatment relieves autistic-like behavioral deficits in three mouse models of autism: Highlights the potential therapeutic applications of sodium bromide in neuropsychiatric disorders, providing insights into its efficacy in alleviating symptoms of autism within experimental settings (Derieux C, Becker JAJ, 2022).

  • Development of Flexible Plasticized Ion Conducting Polymer Blend Electrolytes Based on Polyvinyl Alcohol (PVA): Chitosan (CS) with High Ion Transport Parameters Close to Gel Based Electrolytes: Showcases the role of sodium bromide in the development of high-performance polymer electrolytes for energy storage applications, demonstrating its impact on enhancing ion transport properties (Sadiq NM, Kadir MFZ, 2022).

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vincent Ball
The journal of physical chemistry. B, 123(40), 8405-8410 (2019-09-19)
The gelation kinetics of gelatin depends on the presence of electrolytes, and the influence of cations with variable valency has been recently emphasized. However, acquired knowledge shows that the nature of anions has more influence on various properties of aqueous solutions
Christian Freese et al.
Particle and fibre toxicology, 11, 68-68 (2014-12-30)
In general the prediction of the toxicity and therapeutic efficacy of engineered nanoparticles in humans is initially determined using in vitro static cell culture assays. However, such test systems may not be sufficient for testing nanoparticles intended for intravenous application.
James D Otvos et al.
Clinical chemistry, 61(5), 714-723 (2015-03-18)
Nuclear magnetic resonance (NMR) spectra of serum obtained under quantitative conditions for lipoprotein particle analyses contain additional signals that could potentially serve as useful clinical biomarkers. One of these signals that we named GlycA originates from a subset of glycan
Junji Nemoto et al.
ACS applied materials & interfaces, 7(35), 19809-19815 (2015-08-25)
Simple freeze-drying of 2,2,6,6-tetramethylpiperidine-1-oxyl (TEMPO)-oxidized cellulose nanofibril (TOCN) dispersions in water/tert-butyl alcohol (TBA) mixtures was conducted to prepare TOCN aerogels as high-performance air filter components. The dispersibility of the TOCNs in the water/TBA mixtures, and the specific surface area (SSA)
Alaa S Tulbah et al.
Expert opinion on drug delivery, 12(6), 857-868 (2014-09-23)
This study focuses on the development of a dry powder inhaler (DPI) formulation of simvastatin (SV), and the effects of SV on the respiratory epithelium. Micronised SV samples were prepared by dry jet-milling. The long-term chemical stability and physicochemical properties

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service