Skip to Content
Merck
All Photos(3)

Key Documents

277576

Sigma-Aldrich

Bromine

ACS reagent, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Br2
CAS Number:
Molecular Weight:
159.81
EC Number:
MDL number:
UNSPSC Code:
12141915
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
form:
liquid

grade

ACS reagent

vapor density

7.14 (vs air)

vapor pressure

175 mmHg ( 20 °C)
671 mmHg ( 55 °C)

Assay

≥99.5%

form

liquid

resistivity

7.8E18 μΩ-cm, 20°C

impurities

organic bromine compounds, passes test
≤0.001% I2
≤0.001% S compounds

evapn. residue

≤0.005%

bp

58.8 °C (lit.)

mp

−7.2 °C (lit.)

density

3.119 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.05%

cation traces

Ni: ≤5 ppm
heavy metals: ≤2 ppm

SMILES string

BrBr

InChI

1S/Br2/c1-2

InChI key

GDTBXPJZTBHREO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bromine is a highly volatile and reactive halogen that can be used as a powerful brominating and oxidizing agent in chemical synthesis.

Application

Bromine can be used as a brominating agent for the bromination of:
  • Aromatic and heterocyclic compounds. 
  • Carbonyl compounds at α position to carbonyl groups.
  • Aromatic carboxylic acids via Hell-Volhard-Zelinski reaction in the presence of phosphorus trihalides.
  • Alkylbenzenes at the benzylic position using photocatalytic conditions.

It can also be used as an oxidizing agent for the oxidation of:
  • Primary alcohols to either aldehydes or esters.      
  • Secondary alcohols to ketones.

It can also be used as a Lewis acid catalyst to:     
  • Convert epoxides and CO2 to cyclic carbonates in a continuous flow system.     
  • Synthesize bis(indolyl)methanes by reacting indoles with carbonyl compounds.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reaction of bromine and chlorine with phenolic compounds and natural organic matter extracts-Electrophilic aromatic substitution and oxidation.
Criquet J, et al.
Water Research, 85, 476-486 (2015)
The chemistry of atmospheric bromine.
Wofsy SC, et al.
Geophysical Research Letters, 2(6), 215-218 (1975)
Jennifer A Kozak et al.
Journal of the American Chemical Society, 135(49), 18497-18501 (2013-11-22)
A continuous method for the formation of cyclic carbonates from epoxides and carbon dioxide (CO2) is described. The catalysts used are inexpensive and effective in converting the reagents to the products in a residence time (t(R)) of 30 min. The
Bromine-catalyzed aziridination of olefins. A rare example of atom-transfer redox catalysis by a main group element.
Jeong JUK, et al.
Journal of the American Chemical Society, 120(27), 6844-6845 (1998)
Teresa M Bisson et al.
Environmental science & technology, 46(21), 12186-12193 (2012-10-02)
Recent laboratory and field-scale experiments demonstrated the potential for brominated industrial solid waste from biomass combustion (Br-Ash) to be an efficient, cost-effective alternative to activated carbon for capturing mercury from coal-fired power plants. To develop this attractive alternative technology to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service