Skip to Content
Merck
All Photos(2)

Key Documents

268003

Sigma-Aldrich

Hydrobromic acid

reagent grade, 48%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HBr
CAS Number:
Molecular Weight:
80.91
Beilstein:
3587158
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent grade

grade

reagent grade

Quality Level

vapor density

2.8 (vs air)

vapor pressure

320 psi ( 21.1 °C)
8 mmHg ( 25 °C)

description

48% aqueous HBr

Assay

48%

form

liquid

density

1.49 g/mL at 25 °C (lit.)

SMILES string

Br

InChI

1S/BrH/h1H

InChI key

CPELXLSAUQHCOX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Hydrobromic acid is the strongest mineral acid that is used as a brominating agent in the preparation of organobromine compounds and inorganic bromides. It is also used as a catalyst in a variety of organic reactions.

Application

Hydrobromic acid can be used as:      
  • A catalyst in transesterification reactions.      
  • A reagent to synthesize cellulose nanocrystals (CNCs) via acidic hydrolysis of cotton fibers.     
  • A reactant to prepare denitrosation reagent.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydrobromic acid-catalyzed Friedel-Crafts type reactions of naphthols.
Xu C, et al.
Royal Society of Chemistry Advances, 4(4), 1559-1562 (2014)
The oxidation of acetophenones to arylglyoxals with aqueous hydrobromic acid in dimethyl sulfoxide.
Floyd MB, et al.
The Journal of Organic Chemistry, 50(25), 5022-5027 (1985)
Production of cellulose nanocrystals using hydrobromic acid and click reactions on their surface.
Sadeghifar H, et al.
J. Mater. Sci., 46(22), 7344-7355 (2011)
Efficient bromination of olefins, alkynes, and ketones with dimethyl sulfoxide and hydrobromic acid.
Song S, et al.
Green Chemistry, 17(6), 3285-3289 (2015)
M Y Mirza
Talanta, 27(2), 101-106 (1980-02-01)
The extraction of Pd(II), Rh(III), Ir(III), Au(III) and Pt(IV) from hydrochloric and hydrobromic acid with 5% tri-iso-octylamine solution in carbon tetrachloride has been studied. The gold extract from hydrochloric acid is yellow and absorbs at 325 nm, the palladium compound

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service