221872
Potassium fluoride dihydrate
reagent grade, 98%
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
KF · 2H2O
CAS Number:
Molecular Weight:
94.13
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
grade:
reagent grade
form:
crystals or chunks
Recommended Products
grade
reagent grade
Quality Level
Assay
98%
form
crystals or chunks
mp
41 °C (lit.)
SMILES string
[F-].[K+].[H]O[H].[H]O[H]
InChI
1S/FH.K.2H2O/h1H;;2*1H2/q;+1;;/p-1
InChI key
MZFXHSQFQLKKFD-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Potassium fluoride dihydrate may be used in the following processes:
- As a source of active fluoride ion when used in combination with tetra-n-butylammonium chloride.
- To generate tetrabutylammonium fluoride in situ by reacting with tetrabutylammonium hydrogensulfate, which can catalyze the esterification of carboxylic acids with alkyl halides and ring-opening of aziridines.
- In combination with chlorotrimethylsilane for the desilylation of O-tert-butyldimethylsilyl ethers.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Chromatographia, 28, 179-179 (1989)
Convenient source of 'naked'fluoride: tetra-n-butylammonium chloride and potassium fluoride dihydrate.
Carpino LA and Sau AC.
Journal of the Chemical Society. Chemical Communications, 11, 514-515 (1979)
Efficient halogenation of unsaturated organoaluminum compounds with sulfonyl halides.
Ramazanov IR, et al.
Russ. J. Org. Chem., 49(3), 321-326 (2013)
Hydrogen Bonding in Potassium Fluoride Dihydrate: A Crystallographic, Spectroscopic, and Theoretical Study.
Preisinger A, et al.
Inorganic Chemistry, 33(21), 4774-4780 (1994)
A Mild and Efficient Desilylation of O-tert-Butyldimethylsilyl Ethers Mediated by Chlorotrimethylsilane and Potassium Fluoride Dihydrate in Acetonitrile
Peng Y & Li WDZ
Synlett, 2006(08), 1165-1168 (2006)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service