Skip to Content
Merck
All Photos(1)

Documents

612871

Supelco

Potassium fluoride solution

NMR reference standard, 0.2 M in water, D2O 10 % (99.9 atom % D), NMR tube size 8 mm × 8 in.

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.24

grade

NMR reference standard
analytical standard

composition

D2O, 10% (99.9 atom % D)

concentration

0.2 M in water

technique(s)

NMR: suitable

NMR tube size

8 mm × 8 in.

format

single component solution

SMILES string

[F-].[K+]

InChI

1S/FH.K/h1H;/q;+1/p-1

InChI key

NROKBHXJSPEDAR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

NMR reference standard (0.2 M in water, D2O 10% (99.9 atom % D)) eliminates the sample preparation uncertainty when evaluating instrument performance and assures proper instrument performance in quantitative applications. These NMR reference standards allow the users to monitor key operational parameters such as pw90, sensitivity, resolution, and line shape.

Quantity

8 mm O.D. tube contains 1.9 mL.
Each pack contains 10 tubes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service