Skip to Content
Merck
All Photos(4)

Documents

221376

Sigma-Aldrich

Zinc sulfate heptahydrate

ACS reagent, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnSO4 · 7H2O
CAS Number:
Molecular Weight:
287.56
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Agency

suitable for SM 5210

Assay

99%
99.0-103.0% (ACS specification)

form

crystals or chunks

reaction suitability

reagent type: catalyst
core: zinc

impurities

≤0.01% insolubles

pH

4.4-6.0 (25 °C, 5%)

solubility

water: soluble 965 g/L at 20 °C

anion traces

chloride (Cl-): ≤5 ppm
nitrate (NO3-): ≤0.002%

cation traces

Ca: ≤0.005%
Fe: ≤0.001%
K: ≤0.01%
Mg: ≤0.005%
Mn: ≤3 ppm
NH4+: ≤0.001%
Na: ≤0.05%
Pb: ≤0.003%

SMILES string

O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O

InChI

1S/H2O4S.7H2O.Zn/c1-5(2,3)4;;;;;;;;/h(H2,1,2,3,4);7*1H2;/q;;;;;;;;+2/p-2

InChI key

RZLVQBNCHSJZPX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Zinc sulfate heptahydrate may be used to prepare:
  • Zinc tris(thiourea) sulphate (ZTS), a semiorganic nonlinear optic (NLO) material that may be useful in laser fusion experiments and in second-harmonic-generation (SHG) devices.
  • Highly pure zinc oxide nanostructures via a precipitation method for use in catalytic reactions.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, characterization and optical properties of zinc oxide nanoparticles.
Kumar SS, et al.
International Nano Letters, 3(1), 1-1 (2013)
Kathryn S Evans et al.
PLoS genetics, 16(11), e1008986-e1008986 (2020-11-12)
Zinc is an essential trace element that acts as a co-factor for many enzymes and transcription factors required for cellular growth and development. Altering intracellular zinc levels can produce dramatic effects ranging from cell proliferation to cell death. To avoid
Growth of zinc thiourea sulfate (ZTS) single crystals:: a potential semiorganic NLO material.
Ushasree PM, et al.
Journal of Crystal Growth, 197(1), 216-220 (1999)
J F Nijsen et al.
Biomaterials, 22(22), 3073-3081 (2001-09-29)
Holmium-loaded PLLA microspheres are useful systems in radioembolization therapy of liver metastases because of their low density, biodegradability and favourable radiation characteristics. Neutron activated Ho-loaded microspheres showed a surprisingly low release of the relatively small holmium complex. In this paper
Organic solvents compromise performance of internal standard (ascomycin) in proficiency testing of mass spectrometry-based assays for tacrolimus.
Kimberly L Napoli
Clinical chemistry, 52(4), 765-766 (2006-04-06)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service