Skip to Content
Merck
All Photos(3)

Documents

Z0635

Sigma-Aldrich

Zinc sulfate heptahydrate

meets USP testing specifications

Synonym(s):

Zinc Sulfate hydrate, Zinc sulphate heptahydrate, ZnSO4

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnSO4 · 7H2O
CAS Number:
Molecular Weight:
287.56
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
meets USP testing specifications

Quality Level

Assay

>99.0%

solubility

H2O: 965 g/L at 20 °C

application(s)

pharmaceutical (small molecule)

SMILES string

O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O

InChI

1S/H2O4S.7H2O.Zn/c1-5(2,3)4;;;;;;;;/h(H2,1,2,3,4);7*1H2;/q;;;;;;;;+2/p-2

InChI key

RZLVQBNCHSJZPX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thiago B Kirsten et al.
PloS one, 10(3), e0120263-e0120263 (2015-03-17)
Sickness behavior is considered part of the specific beneficial adaptive behavioral and neuroimmune changes that occur in individuals in response to infectious/inflammatory processes. However, in dangerous and stressful situations, sickness behavior should be momentarily abrogated to prioritize survival behaviors, such
Akira Masuda et al.
Behavioural brain research, 252, 334-338 (2013-06-19)
Social interaction enables animals to transmit various types of sensory information that can modulate learned avoidance behavior and fear responses, which are important to survival. We previously reported that, under a passive avoidance paradigm, avoidance behavior is facilitated when a
Acute diarrhea in adults and children: a global perspective.
Michael Farthing et al.
Journal of clinical gastroenterology, 47(1), 12-20 (2012-12-12)
Kerry J Welsh et al.
American journal of clinical pathology, 140(4), 572-578 (2013-09-21)
To find a suitable method for detecting zinc sulfate in adulterated urine. Two rapid spot tests to detect the presence of zinc sulfate in urine were developed. Addition of 3 to 4 drops of 1N sodium hydroxide solution to approximately
Gaia Dell'Ariccia et al.
The Journal of experimental biology, 216(Pt 8), 1430-1433 (2013-01-12)
Olfactory cues have been shown to be important to homing petrels at night, but apparently those procellariiform species that also come back to the colony during the day are not impaired by smell deprivation. However, the nycthemeral distribution of homing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service