Skip to Content
Merck
All Photos(2)

Key Documents

204986

Sigma-Aldrich

Zinc sulfate heptahydrate

≥99.95% trace metals basis

Synonym(s):

Zinc(II) sulfate heptahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnSO4 · 7H2O
CAS Number:
Molecular Weight:
287.56
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder, crystals or chunks
solubility:
H2O: 965 g/L at 20 °C

grade

for analytical purposes

Quality Level

Agency

suitable for SM 5210

Assay

≥99.95% trace metals basis

form

powder, crystals or chunks

impurities

≤500.0 ppm Trace Metal Analysis

solubility

H2O: 965 g/L at 20 °C

SMILES string

O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O

InChI

1S/H2O4S.7H2O.Zn/c1-5(2,3)4;;;;;;;;/h(H2,1,2,3,4);7*1H2;/q;;;;;;;;+2/p-2

InChI key

RZLVQBNCHSJZPX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Zinc sulfate heptahydrate is a white crystalline solid commonly used as an electrolyte additive in batteries, particularly in zinc-carbon batteries. Additionally, it can be used as a corrosion inhibitor in batteries and a precursor to synthesize nanoparticles.

Application

Zinc sulfate heptahydrate can be used:
  • As an electrolyte for rechargeable hybrid aqueous batteries.
  • As a precursor to synthesize ZnO nanoparticles.
  • As a cathodic corrosion inhibitor in batteries.
Zinc sulfate heptahydrate coated activated carbon can be used in water filtration.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient, One-Step Mechanochemical Process for the Synthesis of ZnO Nanoparticles
Jun Lu, et al.
Industrial & Engineering Chemistry Research, 47, 1095-1101 (2008)
Mechanism of Inhibition of Galvanic Corrosion Between Carbon Steel and Stainless Steel by Sodium N-Lauroyl Sarcosinate and Zinc Sulfate
Shuai Hu, et al.
Surface Review and Letters, 27, 1950146-1950146 (2020)
Zinc-sulphate-heptahydrate coated activated carbon for microbe removal from stormwater
Robert M Guest, et al.
Water Science and Technology, 66, 1582-1589 (2012)
Zinc sulfate treatment of vocal process granuloma.
Taner Yilmaz
European archives of oto-rhino-laryngology : official journal of the European Federation of Oto-Rhino-Laryngological Societies (EUFOS) : affiliated with the German Society for Oto-Rhino-Laryngology - Head and Neck Surgery, 270(3), 1175-1175 (2012-10-12)
Mahmoud Hussein Hadwan et al.
BMC urology, 12, 32-32 (2012-11-14)
Zinc in human seminal plasma is divided into three types of ligands which are high (HMW), intermediate (IMW), and low molecular weight ligands (LMW). The present study was aimed to study the effect of Zn supplementation on the quantitative and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service