Skip to Content
Merck

Skip To

83265

Zinc sulfate solution

BioUltra, Molecular Biology, 2.0 M in H2O

Synonym(s):

Zinc sulfate solution, Zinc Sulfate hydrate, ZnSO4

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
ZnSO4
CAS Number:
Molecular Weight:
161.45
NACRES:
NA.26
PubChem Substance ID:
UNSPSC Code:
12352302
MDL number:
Grade:
Molecular Biology
Form:
solution
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Molecular Biology

Quality Level

product line

BioUltra

form

solution

concentration

2.0 M in H2O

impurities

DNases, none detected, RNases, none detected, phosphatases, none detected, proteases, none detected

pH

4.0±0.5

density

1.31 g/mL at 20 °C

λ

neat

UV absorption

λ: 260 nm Amax: <0.02, λ: 280 nm Amax: <0.02

suitability

passes test for filter test

SMILES string

[Zn++].[O-]S([O-])(=O)=O

InChI

1S/H2O4S.Zn/c1-5(2,3)4;/h(H2,1,2,3,4);/q;+2/p-2

InChI key

NWONKYPBYAMBJT-UHFFFAOYSA-L

Application

Zinc sulfate solution has been used as a component in media for in vitro hyphal growth assays. It has also been used as a supplement in the media along with isopropyl β-D-1-thiogalactopyranoside (IPTG), while expressing zinc containing proteins.

Biochem/physiol Actions

Zinc sulfate solution is as potent as formalin. This chemical is mainly used to treat footrot. It may also be used to treat acute bronchiolitis.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
Z287683266M3409
form

solution

form

aqueous solution

form

liquid

form

liquid

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

200

grade

Molecular Biology

grade

-

grade

Molecular Biology

grade

Molecular Biology

pH

4.0±0.5

pH

-

pH

6.5-8.5 (25 °C)

pH

-

impurities

DNases, none detected, phosphatases, none detected, RNases, none detected, proteases, none detected

impurities

-

impurities

DNases, none detected, RNases, none detected, insoluble matter, passes filter test, phosphatases, none detected, proteases, none detected, <0.002% total nitrogen (N)

impurities

-

density

1.31 g/mL at 20 °C

density

-

density

-

density

-


Still not finding the right product?

Explore all of our products under Zinc sulfate solution


pictograms

CorrosionEnvironment

signalword

Danger

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Diseases of the Musculoskeletal System in the Elderly
Christos I, et al.
Journal of pharmaceutics, 3, 58-62 (2015)
MARCC (Matrix-Assisted Reader Chromatin Capture): An Antibody-Free Method to Enrich and Analyze Combinatorial Nucleosome Modifications
Su Z and Denu JM
Current Protocols in Molecular Biology, 111(1), 21-32 (2015)
J M Anchordoquy et al.
Biological trace element research, 192(2), 175-182 (2019-02-07)
Zinc (Zn) is required for normal reproductive performance in cattle. The aim of this study was to evaluate the effect of subcutaneous injection of 400 mg Zn at the beginning of fixed-time artificial insemination (FTAI) on preovulatory follicle and corpus luteum



Global Trade Item Number

SKUGTIN
83265-250ML-F04061838169686
83265-1L-F04061838177384

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters