Skip to Content
Merck
All Photos(4)

Key Documents

206083

Sigma-Aldrich

Chloroplatinic acid hexahydrate

ACS reagent, ≥37.50% Pt basis

Synonym(s):

Hexachloroplatinic acid hexahydrate, Hydrogen hexachloroplatinate(IV) hexahydrate, Platinic chloride hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2PtCl6 · 6H2O
CAS Number:
Molecular Weight:
517.90
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

form

powder or chunks

reaction suitability

reagent type: catalyst
core: platinum

concentration

≥37.5% Pt

solubility

alcohol: soluble

cation traces

alkalies and other metals: ≤0.05%

SMILES string

O.O.O.O.O.O.Cl.Cl.Cl[Pt](Cl)(Cl)Cl

InChI

1S/6ClH.6H2O.Pt/h6*1H;6*1H2;/q;;;;;;;;;;;;+4/p-4

InChI key

PIJUVEPNGATXOD-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Chloroplatinic acid hexahydrate (H2PtCl6.6H2O) is an inorganic platinum salt. Pure quadrupole resonance frequencies of Cl35 in H2PtCl6.6H2O have been evaluated by spectrophotometric investigations.

Application

Chloroplatinic acid hexahydrate has been used in the following studies:
  • Preparation of Pd nanocubes (150nm width) and Pt nanospheres (150nm diameter).
  • To compose the plating solution for the electrodeposition of Pt nanoparticles on the multilayered graphene petal nanosheets (MGPNs).
  • Synthesis of nano-composites of Pt and nano-carbon (NC).3
Chloroplatinic acid hexahydrate may be used as a platinum precursor in the preparation of ceria supported Pt catalyst, TiO2/Pt nano-composites, Pt nanoparticles impregnated porous carbon films and cadmium sulfide-Pt colloidal system.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemical glutamate biosensing with nanocube and nanosphere augmented single-walled carbon nanotube networks: a comparative study.
Claussen JC, et al.
Journal of Materials Chemistry, 21(30), 11224-11231 (2011)
Synthesis and Characterization of Ceria Supported Platinum via Facile Wet Impregnation from Cerium Based Precursors.
Alam BB, et al.
Journal of Advanced Chemical Engineering, 2015 (2015)
Nanostructuring platinum nanoparticles on multilayered graphene petal nanosheets for electrochemical biosensing.
Claussen JC, et al.
Advances in Functional Materials, 22(16), 3399-3405 (2012)
Photocatalytic properties of TiO2 and TiO2/Pt: A sol-precipitation, sonochemical and hydrothermal approach.
Ultrasonics Sonochemistry, 21(1), 367-375 (2014)
Bolin Chen et al.
ACS applied materials & interfaces, 8(45), 30941-30947 (2016-11-02)
Micro unmanned underwater vehicles (UUVs) need to house propulsion mechanisms that are small in size but sufficiently powerful to deliver on-demand acceleration for tight radius turns, burst-driven docking maneuvers, and low-speed course corrections. Recently, small-scale hydrogen peroxide (H2O2) propulsion mechanisms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service