Skip to Content
Merck
All Photos(3)

Key Documents

13479

Sigma-Aldrich

Sodium thiosulfate pentahydrate

puriss., meets analytical specification of Ph. Eur., BP, USP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2S2O3 · 5H2O
CAS Number:
Molecular Weight:
248.18
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
99.0-100.5% (USP, dry basis)
99.0-101.0% (EP)
form:
crystalline
Pricing and availability is not currently available.

Agency

suitable for EPA 1621
suitable for ISO 25101

Quality Level

grade

puriss.

Assay

99.0-100.5% (USP, dry basis)
99.0-101.0% (EP)

form

crystalline

quality

meets analytical specification of Ph. Eur., BP, USP

impurities

residual solvents, complies
≤0.0005% heavy metals (as Pb)

loss

32-37% loss on drying, 16 h (vacuum)

pH

6.0-8.4 (20 °C, 10%)

anion traces

sulfate, sulfite (as SO42-): ≤2000 mg/kg
sulfide (S2-): in accordance

cation traces

As: ≤3 mg/kg
Ca: ≤50 mg/kg
Fe: ≤10 mg/kg

suitability

complies for appearance of solution

SMILES string

O.O.O.O.O.[Na+].[Na+].[O-]S([O-])(=O)=S

InChI

1S/2Na.H2O3S2.5H2O/c;;1-5(2,3)4;;;;;/h;;(H2,1,2,3,4);5*1H2/q2*+1;;;;;;/p-2

InChI key

PODWXQQNRWNDGD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fingerprint imaging by scanning electrochemical microscopy.
Zhang M and Girault HH.
Electrochemical Communications, 9(7), 1778-1782 (2007)
Sodium thiosulfate pentahydrate: a redetermination by neutron diffraction.
Lisensky GC and Levy HA.
Acta Crystallographica Section B, Structural Science, Crystal Engineering and Materials, 34(6), 1975-1977 (1978)
Nuclear magnetic resonance in sodium thiosulfate pentahydrate.
Murty CRK and El Saffar ZM.
Acta Crystallographica, 15(6), 536-538 (1962)
Melani Sooriyaarachchi et al.
Metallomics : integrated biometal science, 5(3), 197-207 (2013-02-28)
Cis-Platin (CP) is a remarkably effective Pt-based anticancer drug, but it also exhibits severe toxic side-effects, including nephrotoxicity and ototoxicity. Previous studies conducted with mammalian model organisms have clearly demonstrated that the intravenous administration of N-acetyl-L-cysteine (NAC) or sodium thiosulfate
Brooke Bair et al.
Journal of drugs in dermatology : JDD, 10(9), 1042-1044 (2011-11-05)
Calcinosis cutis is a term used to describe a group of disorders which result in calcium deposits in the skin. These disorders can be separated based on etiology. Sodium thiosulfate has been used to systemically treat calciphylaxis with little to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service