Skip to Content
Merck
All Photos(2)

Key Documents

563188

Sigma-Aldrich

Sodium thiosulfate

≥99.99% trace metals basis

Synonym(s):

Disodium thiosulfate, Disodium thiosulphate, Sodium hyposulfite, Sodium oxide sulfide (Na2S2O3 ), Sodium thiosulfate, Sodium thiosulfate (Na2 S2 O3 ), Sodium thiosulfate anhydrous, Sodium thiosulphate, Sulfothiorine, Thiosulfuric acid, disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2S2O3
CAS Number:
Molecular Weight:
158.11
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder
Pricing and availability is not currently available.

grade

for analytical purposes

Assay

≥99.99% trace metals basis

form

powder

impurities

≤100.0 ppm Trace Metal Analysis

SMILES string

[Na+].[Na+].[O-]S([O-])(=O)=S

InChI

1S/2Na.H2O3S2/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

AKHNMLFCWUSKQB-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Sodium thiosulfate is a versatile inorganic compound widely used as a reducing agent and sulfur source to synthesize various nanomaterials. It can also be used in the field of rechargeable batteries, medicine, water treatment, and photography.[1] [2]

Application

Sodium thiosulfate can be used:
  • As a precursor to synthesize gold and silver nanoparticles.[1] [2]
  • As a sulfur source to prepare copper sulfide nanoparticles.[3]
  • As a sulfur passivating agent to prepare highly fluorescent carbon dots for sensing applications.[4]
  • As a dopant and activating agent to fabricate carbon cathodes for Lithium-Sulfur batteries.[5]

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characteristics of copper sulfide nanoparticles obtained in the copper sulfate--sodium thiosulfate system
K S Murasheva, et al
Journal of Structural Chemistry, 58, 1383-1390 (2017)
Tunability and stability of gold nanoparticles obtained from chloroauric acid and sodium thiosulfate reaction
Guandong Zhang, et al.
Nanoscale Research Letters, 7, 1-9 (2012)
Straightforward synthesis of Sulfur/N, S-codoped carbon cathodes for Lithium-Sulfur batteries
Marta Sevilla, et al.
Scientific Reports, 10, 486-486 (2020)
Sculpturing effect of sodium thiosulfate in shape transformation of silver nanoparticles from triangular nanoprisms to hexagonal nanoplates
Bing Liu, et al.
Journal of Nanoscience and Nanotechnology, 11, 5001-5006 (2011)
The Role of N and S Doping on Photoluminescent Characteristics of Carbon Dots from Palm Bunches for Fluorimetric Sensing of Fe3+ Ion
Aphinan Saengsrichan, et al
International Journal of Molecular Sciences, 23, 5001-5001 (2022)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service