Skip to Content
Merck
All Photos(3)

Documents

S6672

Sigma-Aldrich

Sodium thiosulfate pentahydrate

BioXtra, ≥99.5%

Synonym(s):

Disodium thiosulfate pentahydrate, Na2S2O3 · 5H2O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2S2O3 · 5H2O
CAS Number:
Molecular Weight:
248.18
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.25

product line

BioXtra

Quality Level

Assay

≥99.5%

impurities

≤0.0005% Phosphorus (P)
≤0.005% Insoluble matter

solubility

H2O: 1 M, clear, colorless

anion traces

sulfate (SO42-): ≤0.2%

cation traces

Al: ≤0.0005%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

O.O.O.O.O.[Na+].[Na+].[O-]S([O-])(=O)=S

InChI

1S/2Na.H2O3S2.5H2O/c;;1-5(2,3)4;;;;;/h;;(H2,1,2,3,4);5*1H2/q2*+1;;;;;;/p-2

InChI key

PODWXQQNRWNDGD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application


  • Preparation and characterization of sodium thiosulfate pentahydrate/expanded graphite phase change energy storage composites.: The research addresses issues such as high supercooling and phase separation in sodium thiosulfate pentahydrate by adding expanded graphite, significantly improving its thermal properties for energy storage applications (Lu et al., 2020).

  • Surface free energy analysis for stable supercooling of sodium thiosulfate pentahydrate with microcosmic-visualized methods.: This paper presents a detailed analysis of supercooling stability in sodium thiosulfate pentahydrate, employing microcosmic-visualized methods to enhance its application in thermal energy storage systems (Peng et al., 2020).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jeremy A Herrera et al.
Clinical proteomics, 17, 24-24 (2020-06-23)
Haematoxylin and eosin (H&E)-which respectively stain nuclei blue and other cellular and stromal material pink-are routinely used for clinical diagnosis based on the identification of morphological features. A richer characterization can be achieved by laser capture microdissection coupled to mass
Christian Pfeifer et al.
International orthopaedics, 36(7), 1493-1499 (2012-01-13)
The purpose of this study was to find a suitable method of labelling cartilage samples for the measurement of distraction distances in biomechanical testing. Samples of bovine cartilage were labelled using five different methods: hydroquinone and silver nitrate (AgNO3), potassium
Melani Sooriyaarachchi et al.
Metallomics : integrated biometal science, 5(3), 197-207 (2013-02-28)
Cis-Platin (CP) is a remarkably effective Pt-based anticancer drug, but it also exhibits severe toxic side-effects, including nephrotoxicity and ototoxicity. Previous studies conducted with mammalian model organisms have clearly demonstrated that the intravenous administration of N-acetyl-L-cysteine (NAC) or sodium thiosulfate
Michael C Reade et al.
Emergency medicine Australasia : EMA, 24(3), 225-238 (2012-06-08)
Cyanide poisoning is uncommon, but generates interest because of the presumed utility of an antidote immediately available in those areas with a high risk of cyanide exposure. As part of its regular review of guidelines, the Australian Resuscitation Council conducted
Brooke Bair et al.
Journal of drugs in dermatology : JDD, 10(9), 1042-1044 (2011-11-05)
Calcinosis cutis is a term used to describe a group of disorders which result in calcium deposits in the skin. These disorders can be separated based on etiology. Sodium thiosulfate has been used to systemically treat calciphylaxis with little to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service