909173
Sodium thiosulfate
free-flowing, Redi-Dri™, ReagentPlus®, 99%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
Na2S2O3
CAS Number:
Molecular Weight:
158.11
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21
Assay:
99%
grade:
anhydrous
form:
powder
Recommended Products
grade
anhydrous
Agency
suitable for EPA 601
product line
ReagentPlus®
Redi-Dri™
Assay
99%
form
powder
quality
free-flowing
pH
6-8.5 (20 °C, 50 g/L)
SMILES string
[Na+].[Na+].[O-]S([O-])(=O)=S
InChI
1S/2Na.H2O3S2/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2
InChI key
AKHNMLFCWUSKQB-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
General description
Sodium thiosulfate (Na2S2O3) can be obtained from Na2S2O3.10H2O by heating above 100 °C. It can be synthesized from sodium sulfate.
Application
Applied in a widely useful, thiosulfate-assisted, synthesis of a variety of diamine-CuCN complexes.
Sodium thiosulfate (Na2S2O3) may be used in the following studies:
Sodium thiosulfate (Na2S2O3) may be used in the following studies:
- Industrial synthesis of methylene blue.
- Quantitative estimation of elemental iodine by titrimetry.
- Detection of trace levels of cysteine by employing triangular silver nanoplates (TAg-NPs).
Redi-Dri™ is an innovative packaging product line that helps eliminate clumping material by adding extra packaging process steps without any chemical additives, anti-clumping agents, or hydrophobic compounds. Redi Dri offers a more convenient and safer way to handle the products.
Legal Information
ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Redi-Dri is a trademark of Sigma-Aldrich Co. LLC
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service