Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
1.02790
Copper(II) sulfate pentahydrate
for analysis EMSURE® ACS,ISO,Reag. Ph Eur
Synonym(s):
Copper(II) sulfate pentahydrate
About This Item
grade
ACS reagent
Quality Level
Agency
reag. ISO
reag. Ph. Eur.
product line
EMSURE®
Assay
99.0-100.5% (iodometric)
form
solid
potency
960 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)
impurities
≤0.005% Insoluble matter
pH
3.5-4.5 (20 °C, 50 g/L in H2O)
mp
110 °C (230 °F)
density
2.284 g/cm3 at 20 °C
anion traces
chloride (Cl-): ≤0.0005%
cation traces
Ca: ≤0.005%
Fe: ≤0.003%
K: ≤0.001%
N: ≤0.001%
Na: ≤0.005%
Ni: ≤0.005%
Pb: ≤0.005%
Zn: ≤0.03%
storage temp.
2-30°C
SMILES string
[Cu+2].[S](=O)(=O)([O-])[O-].O.O.O.O.O
InChI
1S/Cu.H2O4S.5H2O/c;1-5(2,3)4;;;;;/h;(H2,1,2,3,4);5*1H2/q+2;;;;;;/p-2
InChI key
JZCCFEFSEZPSOG-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
- Synthesis and antifungal activity of slow-release pyridinylcarbonyl chitooligosaccharide copper complexes.: This study explores the synthesis of slow-release copper complexes with antifungal properties, highlighting their potential applications in agricultural and medical settings (Liu et al., 2022).
Linkage
Analysis Note
Insoluble matter: ≤ 0.005 %
Chloride (Cl): ≤ 0.0005 %
Total nitrogen (N): ≤ 0.001 %
Ca (Calcium): ≤ 0.005 %
Fe (Iron): ≤ 0.003 %
K (Potassium): ≤ 0.001 %
Na (Sodium): ≤ 0.005 %
Ni (Nickel): ≤ 0.005 %
Pb (Lead): ≤ 0.005 %
Zn (Zinc): ≤ 0.03 %
Corresponds to ACS,ISO,Reag. Ph Eur
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Helpful?
-
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdfHelpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service