Skip to Content
Merck
All Photos(2)

Key Documents

T8127

Sigma-Aldrich

Glyceryl tripalmitate

≥85%

Synonym(s):

TG(16:0/16:0/16:0), Glycerol tripalmitate, Tripalmitin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)14COOCH2]2CHOCO(CH2)14CH3
CAS Number:
Molecular Weight:
807.32
Beilstein:
1811188
EC Number:
MDL number:
UNSPSC Code:
41141833
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Assay

≥85%

form

powder

functional group

ester

lipid type

neutral glycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCC

InChI

1S/C51H98O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-49(52)55-46-48(57-51(54)45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)47-56-50(53)44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h48H,4-47H2,1-3H3

InChI key

PVNIQBQSYATKKL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao-Li Qin et al.
Journal of agricultural and food chemistry, 60(9), 2377-2384 (2012-03-01)
Tripalmitin-enriched triacylglycerols were concentrated from palm stearin by acetone fractionation and as the substrate reacted with a mixture of equimolar quantities of fatty acids (C8:0-C18:3). The incorporation degree and acyl migration level of the fatty acids and acylglycerols composition were
Maike Windbergs et al.
AAPS PharmSciTech, 11(1), 2-8 (2010-03-24)
Extrudates based on varying ratios of the triglyceride tripalmitin and the hydrophilic polymer polyethylene glycol as matrix formers were produced as oral dosage forms with controlled release characteristics. The extrudates were processed below the melting points of the excipients and
Ming-Jun Tsai et al.
International journal of pharmaceutics, 423(2), 461-470 (2011-12-24)
The objective of the present work was to investigate the specific brain targeting of baicalein by intravenous injection after incorporation into nanostructured lipid carriers (NLCs). The NLC system, composed of tripalmitin, Gelucires, vitamin E, phospholipids, and poloxamer 188 (referred to
Maximilian Greiner et al.
Journal of agricultural and food chemistry, 60(20), 5243-5249 (2012-04-17)
Using molecular-dynamics (MD) simulations the densities and self-diffusion coefficients of a range of liquid monoacid triacylglycerides (TAGs) have been studied as a function of temperature and, for the first time, pressure. While offset by their ambient properties, the response of
Rieke Witzleb et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 77(1), 170-177 (2010-10-14)
The aim of this work was to develop a continuous solid lipid extrusion process that includes post-process milling of the extrudates. Die diameters smaller than 0.5 mm should be used for taste masking of the bitter tasting anthelmintic praziquantel. During

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service