Skip to Content
Merck
All Photos(1)

Key Documents

PHR1729

Supelco

Buprenorphine Hydrochloride

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Buprenorphine hydrochloride, [5α,7α(S)]-17-(Cyclopropylmethyl)-α-(1,1-dimethylethyl)4,5-epoxy-18,19-dihydo-3-hydroxy-6-methoxy-α-methyl-6,14-ethenomorphinan-7-methanol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H41NO4 · HCl
CAS Number:
Molecular Weight:
504.10
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0001109
traceable to USP 1078700

API family

buprenorphine

CofA

current certificate can be downloaded

packaging

pkg of 50 mg

drug control

USDEA Schedule III; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IIC (Portugal)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

Cl[H].CO[C@]12CC[C@@]3(C[C@@H]1[C@](C)(O)C(C)(C)C)[C@H]4Cc5ccc(O)c6O[C@@H]2[C@]3(CCN4CC7CC7)c56

InChI

1S/C29H41NO4.ClH/c1-25(2,3)26(4,32)20-15-27-10-11-29(20,33-5)24-28(27)12-13-30(16-17-6-7-17)21(27)14-18-8-9-19(31)23(34-24)22(18)28;/h8-9,17,20-21,24,31-32H,6-7,10-16H2,1-5H3;1H/t20-,21-,24-,26+,27-,28+,29-;/m1./s1

InChI key

UAIXRPCCYXNJMQ-RZIPZOSSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certified pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Buprenorphine Hydrochloride is a potent long-acting synthetic opiate analgesic.[1]

Application

Buprenorphine Hydrochloride may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by high performance liquid chromatography method.[2]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Agonist at ORL1 receptors; antagonist or partial agonist at μ, κ, and δ opioid receptors.
Full agonist at ORL1 receptors, antagonist or partial agonist at μ, κ, and δ opioid receptors.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA7438 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and validation of a HPLC method for the determination of buprenorphine hydrochloride, naloxone hydrochloride and noroxymorphone in a tablet formulation
Mostafavi A, et al.
Talanta, 77(4), 1415-1419 (2009)
The respiratory depressive effects of intravenous buprenorphine in patients in an intensive care unit.
Downing JW, et al.
South African Medical Journal = Suid-Afrikaanse Tydskrif Vir Geneeskunde, 55(25), 1023-1027 (1979)
Liqi Shen et al.
American journal of translational research, 13(1), 24-37 (2021-02-03)
Bone fracture induces an acute inflammatory response in the resident and peripheral monocyte/macrophage cells. Excessive amounts of proinflammatory cytokines can cause severe tissue damage and inhibit bone healing. The proinflammatory cytokine genes are mainly controlled by TLR4/NF-κB (Toll-like receptor 4/Nuclear

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service