Skip to Content
Merck
All Photos(1)

Key Documents

B7536

Supelco

Buprenorphine hydrochloride solution

analytical standard, for drug analysis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H41NO4 · HCl
CAS Number:
Molecular Weight:
504.10
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard, for drug analysis

Quality Level

drug control

Home Office Schedule 3; psychotrope (France); kontrollierte Droge in Deutschland; (Spain); Decreto Lei 15/93: Tabela IIC (Portugal)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

−20°C

SMILES string

Cl.CO[C@]12CC[C@@]3(CC1[C@](C)(O)C(C)(C)C)[C@H]4Cc5ccc(O)c6OC2[C@]3(CCN4CC7CC7)c56

InChI

1S/C29H41NO4.ClH/c1-25(2,3)26(4,32)20-15-27-10-11-29(20,33-5)24-28(27)12-13-30(16-17-6-7-17)21(27)14-18-8-9-19(31)23(34-24)22(18)28;/h8-9,17,20-21,24,31-32H,6-7,10-16H2,1-5H3;1H/t20-,21-,24-,26+,27-,28+,29-;/m1./s1

InChI key

UAIXRPCCYXNJMQ-RZIPZOSSSA-N

Application

Buprenorphine hydrochloride solution may be used as a reference standard in the determination of buprenorphine in samples of human plasma using gas chromatography coupled with mass spectrometry (GC/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bilin Liu et al.
STAR protocols, 1(3), 100186-100186 (2020-12-31)
Here, we present a modified protocol for a mouse heart failure (HF) model using minimally invasive transverse aortic constriction (miTAC). miTAC is a more effective method in mice than the standard open-chest transverse aortic constriction (TAC) to generate an HF
Development and validation of a sensitive analytical method for the simultaneous determination of buprenorphine and norbuprenorphine in human plasma.
Gopal S, et al.
European Journal of Pharmaceutics and Biopharmaceutics, 51(2), 147-151 (2001)
Afarin Rahimi-Movaghar et al.
The Cochrane database of systematic reviews, 1(1), CD007775-CD007775 (2013-02-27)
Pharmacologic therapies for maintenance treatment of heroin dependence have been used and studied widely. Systematic reviews have demonstrated the effectiveness of such therapies. Opium dependence is associated with less problems and impairments and is less likely to be used by
Robert P Schwartz et al.
American journal of public health, 103(5), 917-922 (2013-03-16)
We examined the association between the expansion of methadone and buprenorphine treatment and the prevalence of heroin overdose deaths in Baltimore, Maryland from 1995 to 2009. We conducted a longitudinal time series analysis of archival data using linear regression with
Karen L Cropsey et al.
Journal of addiction medicine, 7(3), 210-215 (2013-04-24)
This project sought to demonstrate the feasibility and acceptability of providing on-site buprenorphine treatment to individuals under community corrections supervision. Seventeen women and 13 men were enrolled on-site over a 2-week period at a community corrections location. Study participants received

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service