Skip to Content
Merck
All Photos(1)

Key Documents

SML0711

Sigma-Aldrich

RS504393

≥98% (HPLC)

Synonym(s):

6-Methyl-1′-[2-(5-methyl-2-phenyl-4-oxazolyl)ethyl]-spiro[4H-3,1-benzoxazine-4,4′-piperidin]-2(1H)-one, RS-504393

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H27N3O3
CAS Number:
Molecular Weight:
417.50
UNSPSC Code:
51111800
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 1 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

N3(CCC4(OC(=O)Nc5c4cc(cc5)C)CC3)CCc1nc([o]c1C)c2ccccc2

InChI

1S/C25H27N3O3/c1-17-8-9-22-20(16-17)25(31-24(29)27-22)11-14-28(15-12-25)13-10-21-18(2)30-23(26-21)19-6-4-3-5-7-19/h3-9,16H,10-15H2,1-2H3,(H,27,29)

InChI key

ODNICNWASXKNNQ-UHFFFAOYSA-N

Application

RS504393 has been used in mass spectrometry.[1]

Biochem/physiol Actions

RS504393 helps to increase the analgesic potency of morphine and buprenorphine in neuropathic rats. RS504393 has the ability to block the upregulation of pronociceptive factors, like IL-1 (interlukin 1) β, IL-18 (interlukin 18), IL-6 (interlukin 6) and inducible nitric oxide synthase (iNOS), that are induced by chronic constriction injury (CCI).[2]
RS504393 is a highly selective CCR2 chemokine receptor antagonist.
RS504393 is a highly selective CCR2 chemokine receptor antagonist. RS504393 inhibits MCP-1 chemotaxis and ischemia-reperfusion injury in kidneys.

Features and Benefits

This compound is featured on the Chemokine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The RS504393 influences the level of nociceptive factors and enhances opioid analgesic potency in neuropathic rats
Kwiatkowski K, et al.
Journal of Neuroimmune Pharmacology, 12(3), 402-419 (2017)
Chrono-pharmacological Targeting of the CCL2-CCR2 Axis Ameliorates Atherosclerosis
Winter C, et al.
Cell Metabolism (2018)
Bradley W Richmond et al.
Mucosal immunology, 14(2), 431-442 (2020-09-25)
Although activation of adaptive immunity is a common pathological feature of chronic obstructive pulmonary disease (COPD), particularly during later stages of the disease, the underlying mechanisms are poorly understood. In small airways of COPD patients, we found that localized disruption
Maira Licia Foresti et al.
Epilepsy & behavior : E&B, 102, 106695-106695 (2019-12-01)
Neuroinflammation role on epileptogenesis has been the subject of increasing interest. Many studies showed elevation in cytokines and chemokines expression following seizures, such as, CCL2 protein (C-C motif ligand 2 chemokine) and its specific receptor, CCR2. In addition, recent studies

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service