Skip to Content
Merck
All Photos(1)

Key Documents

P8155

Sigma-Aldrich

(R)-(−)-Phenylephrine hydrochloride

analytical standard

Synonym(s):

(R)-(−)-1-(3-Hydroxyphenyl)-2-methylaminoethanol hydrochloride, (R)-(−)-3-Hydroxy-α-(methylaminomethyl)benzyl alcohol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CH(CH2NHCH3)OH·HCl
CAS Number:
Molecular Weight:
203.67
Beilstein:
4158948
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

grade

analytical standard

Quality Level

form

powder

packaging

vial of 250 mg

mp

143-145 °C (lit.)

SMILES string

OC1=CC([C@H](CNC)O)=CC=C1.Cl

InChI

1S/C9H13NO2.ClH/c1-10-6-9(12)7-3-2-4-8(11)5-7;/h2-5,9-12H,6H2,1H3;1H/t9-;/m0./s1

InChI key

OCYSGIYOVXAGKQ-FVGYRXGTSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

α-Adrenergic agonist

Packaging

Supplied in amber screw-cap vials

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Edmund Tsui et al.
Experimental eye research, 112, 134-138 (2013-05-25)
The impact of mydriatic agents on the standardized provocation of retinal vascular reactivity has not been systematically investigated. Our aim was to investigate the effect of commonly used mydriatic agents on the provoked vascular response of retinal arterioles. One eye
Thomas R Sundermeier et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 28(8), 3780-3791 (2014-05-09)
Photoreceptor cell death is the proximal cause of blindness in many retinal degenerative disorders; hence, understanding the gene regulatory networks that promote photoreceptor survival is at the forefront of efforts to combat blindness. Down-regulation of the microRNA (miRNA)-processing enzyme DICER1
Doreen Schmidl et al.
Investigative ophthalmology & visual science, 54(3), 1964-1970 (2013-02-27)
We determined whether administration of a nitric oxide synthase (NOS) inhibitor alters optic nerve head blood flow (ONHBF) regulation during isometric exercise in healthy subjects. Our study was done in a randomized, placebo-controlled, double-masked, three-way crossover design. A total of
Dale A Currigan et al.
Anesthesiology, 121(5), 930-936 (2014-09-10)
Vasopressor drugs, commonly used to treat systemic hypotension and maintain organ perfusion, may also induce regional vasoconstriction in specialized vascular beds such as the lung. An increase in pulmonary vascular tone may adversely affect patients with pulmonary hypertension or right
C A Boly et al.
British journal of anaesthesia, 111(5), 750-758 (2013-07-11)
Left-ventricular end-systolic elastance (Ees) is an index of cardiac contractility, but the invasive nature of its assessment has limited perioperative application. We explored the feasibility of a minimally invasive method of Ees estimation for perioperative assessment of cardiac function and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service