Merck
  • DICER1 is essential for survival of postmitotic rod photoreceptor cells in mice.
DICER1 is essential for survival of postmitotic rod photoreceptor cells in mice.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2014-05-09)
Thomas R Sundermeier, Ning Zhang, Frans Vinberg, Debarshi Mustafi, Hideo Kohno, Marcin Golczak, Xiaodong Bai, Akiko Maeda, Vladimir J Kefalov, Krzysztof Palczewski
ABSTRACT
Photoreceptor cell death is the proximal cause of blindness in many retinal degenerative disorders; hence, understanding the gene regulatory networks that promote photoreceptor survival is at the forefront of efforts to combat blindness. Down-regulation of the microRNA (miRNA)-processing enzyme DICER1 in the retinal pigmented epithelium has been implicated in geographic atrophy, an advanced form of age-related macular degeneration (AMD). However, little is known about the function of DICER1 in mature rod photoreceptor cells, another retinal cell type that is severely affected in AMD. Using a conditional-knockout (cKO) mouse model, we report that loss of DICER1 in mature postmitotic rods leads to robust retinal degeneration accompanied by loss of visual function. At 14 wk of age, cKO mice exhibit a 90% reduction in photoreceptor nuclei and a 97% reduction in visual chromophore compared with those in control littermates. Before degeneration, cKO mice do not exhibit significant defects in either phototransduction or the visual cycle, suggesting that miRNAs play a primary role in rod photoreceptor survival. Using comparative small RNA sequencing analysis, we identified rod photoreceptor miRNAs of the miR-22, miR-26, miR-30, miR-92, miR-124, and let-7 families as potential factors involved in regulating the survival of rods.
Sigma-Aldrich
(R)-(−)-Phenylephrine hydrochloride, analytical standard
Sigma-Aldrich
(R)-(−)-Phenylephrine hydrochloride, powder
Phenylephrine hydrochloride for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Phenylephrine Hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Xylazine, ≥99%
USP
Phenylephrine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Phenylephrine hydrochloride, European Pharmacopoeia (EP) Reference Standard